Gene Transcripts

Gene Symbol
OR5L1
Entrez Gene ID
219437
Gene Name
olfactory receptor family 5 subfamily L member 1 (gene/pseudogene)
Gene Aliases
OR11-151, OST262
Chromosome Location
Chr. 11: 55811467 - 55812402 on Build GRCh38
UniGene ID
Hs.553623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004738.1 NP_001004738.1 1 324
GenBank mRNA BC109249.1 - 1 324
BC109250.1 - 1 324 40022959
BC136939.1 - 1 424 9020931

View Full Product Details