Gene Transcripts

Gene Symbol
OR5B17
Entrez Gene ID
219965
Gene Name
olfactory receptor family 5 subfamily B member 17
Gene Aliases
OR11-237, OR5B20P
Chromosome Location
Chr. 11: 58358125 - 58359069 on Build GRCh38
UniGene ID
Hs.554517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005489.1 NP_001005489.1 1 507

View Full Product Details