Gene Transcripts
- Gene Symbol
- TMEM2
- Entrez Gene ID
- 23670
- Gene Name
- transmembrane protein 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 71683366 - 71769532 on Build GRCh38
- UniGene ID
- Hs.494146
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135820.1 |
NP_001129292.1
|
9, 10 |
2408 |
|
NM_013390.2 |
NP_037522.1
|
10, 11 |
2597 |
|
XM_005251869.4 |
XP_005251926.1
|
10, 11 |
2230 |
|
GenBank mRNA |
AB037833.2 |
- |
10, 11 |
2190 |
|
AF137030.1 |
- |
10, 11 |
2205 |
|
AK075370.1 |
- |
3, 4 |
325 |
|
BC140773.1 |
- |
9, 10 |
1910 |
9021641 |
BC144510.1 |
- |
10, 11 |
2065 |
|
BC146780.1 |
- |
10, 11 |
2190 |
9007233 |
BC171782.1 |
- |
9, 10 |
1911 |
|
View Full Product Details