Gene Transcripts
- Gene Symbol
- FUT2
- Entrez Gene ID
- 2524
- Gene Name
- fucosyltransferase 2
- Gene Aliases
- B12QTL1, SE, SEC2, Se2, sej
- Chromosome Location
- Chr. 19: 48695971 - 48705934 on Build GRCh38
- UniGene ID
- Hs.579928
- Species
- Homo sapiens
- Gene Symbol
- LOC105447645
- Entrez Gene ID
- 105447645
- Gene Name
- uncharacterized LOC105447645
- Gene Aliases
- Chromosome Location
- Chr. 19: 48702450 - 48703848 on Build GRCh38
- UniGene ID
- Hs.655052
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_131188.1 |
|
1 |
817 |
|
GenBank mRNA |
BC080570.1 |
- |
1 |
817 |
|
View Full Product Details