Gene Transcripts
- Gene Symbol
- OSTF1
- Entrez Gene ID
- 26578
- Gene Name
- osteoclast stimulating factor 1
- Gene Aliases
- OSF, SH3P2, bA235O14.1
- Chromosome Location
- Chr. 9: 75088480 - 75147201 on Build GRCh38
- UniGene ID
- Hs.494192
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012383.4 |
NP_036515.4
|
6, 7 |
573 |
|
XM_006717053.3 |
XP_006717116.1
|
6, 7 |
582 |
|
XM_006717055.3 |
XP_006717118.1
|
5, 6 |
525 |
|
XM_011518524.2 |
XP_011516826.1
|
7, 8 |
392 |
|
XM_011518525.2 |
XP_011516827.1
|
6, 7 |
584 |
|
XM_017014622.1 |
XP_016870111.1
|
6, 7 |
574 |
|
GenBank mRNA |
AK222596.1 |
- |
6, 7 |
517 |
|
AK291511.1 |
- |
6, 7 |
541 |
|
BC007459.1 |
- |
6, 7 |
479 |
4052054 |
U63717.1 |
- |
6, 7 |
511 |
|
View Full Product Details