Gene Transcripts
- Gene Symbol
- NFU1
- Entrez Gene ID
- 27247
- Gene Name
- NFU1 iron-sulfur cluster scaffold
- Gene Aliases
- CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU
- Chromosome Location
- Chr. 2: 69396113 - 69438123 on Build GRCh38
- UniGene ID
- Hs.430439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002755.2 |
NP_001002755.1
|
6, 7 |
755 |
|
NM_001002756.2 |
NP_001002756.1
|
5, 6 |
709 |
|
NM_015700.3 |
NP_056515.2
|
6, 7 |
845 |
|
NR_045631.1 |
|
5, 6 |
619 |
|
XM_017003808.1 |
XP_016859297.1
|
6, 7 |
558 |
|
GenBank mRNA |
AA625581.1 |
- |
2, 3 |
340 |
|
AF132967.1 |
- |
6, 7 |
557 |
|
AI129426.1 |
- |
2, 3 |
349 |
|
AJ132584.1 |
- |
6, 7 |
633 |
|
AK300700.1 |
- |
6, 7 |
564 |
|
AK314004.1 |
- |
6, 7 |
588 |
|
AY286306.1 |
- |
6, 7 |
602 |
|
AY286307.1 |
- |
6, 7 |
692 |
|
AY335194.1 |
- |
6, 7 |
748 |
|
BC113692.1 |
- |
6, 7 |
717 |
8322744 |
BC113694.1 |
- |
6, 7 |
717 |
8322746 |
BG385940.1 |
- |
5, 6 |
507 |
|
BU178674.1 |
- |
5, 6 |
451 |
|
BU507991.1 |
- |
6, 7 |
560 |
|
BX538347.1 |
- |
6, 7 |
620 |
|
View Full Product Details