Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013444.3 | NP_038472.2 | 1 | 466 | |
GenBank mRNA | AF189009.1 | - | 1 | 420 | |
AF293385.1 | - | 1 | 185 | ||
AK297252.1 | - | 1 | 432 | ||
AK302885.1 | - | 1 | 432 | ||
AL442081.1 | - | 1 | 79 | ||
BC069237.1 | - | 2 | 337 | 4543266 | |
BI602998.1 | - | 1 | 474 |