Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000041.3 | NP_000032.1 | 2 | 129 | |
NM_001302688.1 | NP_001289617.1 | 2 | 211 | ||
NM_001302689.1 | NP_001289618.1 | 2 | 60 | ||
NM_001302690.1 | NP_001289619.1 | 2 | 160 | ||
NM_001302691.1 | NP_001289620.1 | 2 | 144 | ||
GenBank mRNA | AK308546.1 | - | 2 | 82 | |
AK314898.1 | - | 2 | 82 | ||
BC003557.1 | - | 2 | 58 | 3355712 | |
BI550631.1 | - | 2 | 106 | ||
BM042032.1 | - | 2 | 144 | ||
BU848796.1 | - | 96 | |||
CA771535.1 | - | 1 | 278 | ||
DB486513.1 | - | 2 | 61 | ||
HY056112.1 | - | 2 | 129 | ||
HY074384.1 | - | 2 | 60 | ||
K00396.1 | - | 2 | 73 | ||
KU177910.1 | - | 1 | 13 | ||
KU177911.1 | - | 1 | 13 | ||
KU177912.1 | - | 1 | 13 | ||
KU177913.1 | - | 1 | 13 | ||
M12529.1 | - | 2 | 74 | ||
Z70760.1 | - | 1 | 50 |