Gene Transcripts
- Gene Symbol
- IGFBP7
- Entrez Gene ID
- 3490
- Gene Name
- insulin like growth factor binding protein 7
- Gene Aliases
- AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF
- Chromosome Location
- Chr. 4: 57031071 - 57110385 on Build GRCh38
- UniGene ID
- Hs.479808
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253835.1 |
NP_001240764.1
|
3, 4 |
734 |
|
NM_001553.2 |
NP_001544.1
|
3, 4 |
734 |
|
GenBank mRNA |
AI761388.1 |
- |
2, 3 |
401 |
|
AK303915.1 |
- |
3, 4 |
736 |
|
AK316082.1 |
- |
3, 4 |
734 |
|
BC017201.2 |
- |
3, 4 |
722 |
3632247 |
BC066339.1 |
- |
3, 4 |
734 |
5296697 |
BT006654.1 |
- |
3, 4 |
700 |
|
L19182.1 |
- |
3, 4 |
713 |
|
S75725.1 |
- |
3, 4 |
722 |
|
View Full Product Details