Gene Transcripts
- Gene Symbol
- MFAP2
- Entrez Gene ID
- 4237
- Gene Name
- microfibrillar associated protein 2
- Gene Aliases
- MAGP, MAGP-1, MAGP1
- Chromosome Location
- Chr. 1: 16974502 - 16981649 on Build GRCh38
- UniGene ID
- Hs.389137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135247.1 |
NP_001128719.1
|
8, 9 |
567 |
|
NM_001135248.1 |
NP_001128720.1
|
8, 9 |
586 |
|
NM_002403.3 |
NP_002394.1
|
8, 9 |
589 |
|
NM_017459.2 |
NP_059453.1
|
8, 9 |
570 |
|
XM_011541476.2 |
XP_011539778.1
|
8, 9 |
1196 |
|
GenBank mRNA |
AK222751.1 |
- |
8, 9 |
548 |
|
BC015039.1 |
- |
8, 9 |
589 |
3910064 |
CR407678.1 |
- |
7, 8 |
456 |
|
DN996554.1 |
- |
8, 9 |
539 |
|
U19718.1 |
- |
7, 8 |
497 |
|
View Full Product Details