Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001035005.3 | NP_001030177.1 | 2 | 357 | |
NM_001199346.1 | NP_001186275.1 | 2 | 193 | ||
GenBank mRNA | AB097012.1 | - | 2 | 277 | |
AK027111.1 | - | 2 | 153 | ||
AK291882.1 | - | 2 | 164 | ||
BC022357.1 | - | 2 | 162 | 4297017 | |
BC093004.1 | - | 2 | 314 | 5273608 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199355.1 | NP_001186284.1 | 6 | 645 | |
NM_001199356.1 | NP_001186285.1 | 6 | 712 | ||
GenBank mRNA | BX393840.2 | - | 6 | 647 |