Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016060.2 | NP_057144.1 | 4 | 414 | |
XM_017024710.1 | XP_016880199.1 | 3 | 487 | ||
GenBank mRNA | AF151883.1 | - | 4 | 386 | |
AK130946.1 | - | 3 | 317 | ||
AK311875.1 | - | 4 | 383 | ||
BC012539.1 | - | 4 | 389 | 4248123 |