Gene Transcripts

Gene Symbol
BOLA1
Entrez Gene ID
51027
Gene Name
bolA family member 1
Gene Aliases
CGI-143
Chromosome Location
Chr. 1: 149899582 - 149900795 on Build GRCh38
UniGene ID
Hs.13880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321025.1 NP_001307954.1 2 413
NM_001321026.1 NP_001307955.1 2 471
NM_016074.4 NP_057158.1 2 246
XM_006711348.3 XP_006711411.1 2 279
GenBank mRNA AF151901.1 - 2 181
AK313014.1 - 1 562
BC002771.2 - 2 206
BC063405.1 - 2 206 5190565
BF970430.1 - 427
CA389141.1 - 2 226
CB992246.1 - 2 389

View Full Product Details