Gene Transcripts

Gene Symbol
PI15
Entrez Gene ID
51050
Gene Name
peptidase inhibitor 15
Gene Aliases
CRISP8, P24TI, P25TI
Chromosome Location
Chr. 8: 74824537 - 74855044 on Build GRCh38
UniGene ID
Hs.98558
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324403.1 NP_001311332.1 6 959
NM_015886.4 NP_056970.1 6 931
GenBank mRNA BC074931.2 - 5 775
BC074932.2 - 5 775 30915266
BC126290.1 - 5 780 8992006
BC126292.1 - 5 782 8992008
CR749657.1 - 6 932
D45027.1 - 5 1236

View Full Product Details