Gene Transcripts

Gene Symbol
ASB1
Entrez Gene ID
51665
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
ASB-1
Chromosome Location
Chr. 2: 238426741 - 238452250 on Build GRCh38
UniGene ID
Hs.516788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040445.1 NP_001035535.1 1, 2 134
XM_005246080.1 XP_005246137.1 1, 2 378
GenBank mRNA AF156777.1 - 1, 2 48
BC014528.2 - 1, 2 134 3842384

View Full Product Details