Gene Transcripts
- Gene Symbol
- BCAS4
- Entrez Gene ID
- 55653
- Gene Name
- breast carcinoma amplified sequence 4
- Gene Aliases
- CNOL
- Chromosome Location
- Chr. 20: 50794894 - 50877736 on Build GRCh38
- UniGene ID
- Hs.381178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001010974.2 |
NP_001010974.1
|
2, 3 |
356 |
|
NM_017843.4 |
NP_060313.3
|
2, 3 |
356 |
|
NM_198799.3 |
NP_942094.2
|
2, 3 |
356 |
|
XM_011528886.2 |
XP_011527188.1
|
2, 3 |
279 |
|
XM_011528887.2 |
XP_011527189.1
|
2, 3 |
279 |
|
XM_017027932.1 |
XP_016883421.1
|
2, 3 |
279 |
|
GenBank mRNA |
AF361220.1 |
- |
2, 3 |
356 |
|
AJ511266.1 |
- |
2, 3 |
180 |
|
AK000502.1 |
- |
2, 3 |
219 |
|
BC038381.1 |
- |
2, 3 |
206 |
4472674 |
BC047337.1 |
- |
2, 3 |
185 |
|
View Full Product Details