Gene Transcripts
- Gene Symbol
- CEP85
- Entrez Gene ID
- 64793
- Gene Name
- centrosomal protein 85
- Gene Aliases
- CCDC21
- Chromosome Location
- Chr. 1: 26234153 - 26278810 on Build GRCh38
- UniGene ID
- Hs.109051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281517.2 |
NP_001268446.1
|
8, 9 |
1674 |
|
NM_001281518.2 |
NP_001268447.1
|
2, 3 |
235 |
|
NM_001319944.1 |
NP_001306873.1
|
9, 10 |
1827 |
|
NM_022778.4 |
NP_073615.2
|
9, 10 |
1827 |
|
XM_017002105.1 |
XP_016857594.1
|
9, 10 |
1827 |
|
GenBank mRNA |
AK024038.1 |
- |
9, 10 |
1783 |
|
AK025653.1 |
- |
3, 4 |
426 |
|
AK299312.1 |
- |
8, 9 |
1627 |
|
AL133609.1 |
- |
5, 6 |
668 |
|
AL137420.1 |
- |
3, 4 |
425 |
|
BC019902.1 |
- |
2, 3 |
228 |
|
BC064528.1 |
- |
9, 10 |
1778 |
5585742 |
DA666742.1 |
- |
2, 3 |
235 |
|
View Full Product Details