Gene Transcripts
- Gene Symbol
- SULT1C2
- Entrez Gene ID
- 6819
- Gene Name
- sulfotransferase family 1C member 2
- Gene Aliases
- ST1C1, ST1C2, SULT1C1, humSULTC2
- Chromosome Location
- Chr. 2: 108288639 - 108309915 on Build GRCh38
- UniGene ID
- Hs.436123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001056.3 |
NP_001047.1
|
4, 5 |
835 |
|
NM_176825.2 |
NP_789795.1
|
5, 6 |
868 |
|
XM_005264013.3 |
XP_005264070.1
|
4, 5 |
656 |
|
GenBank mRNA |
AB008164.1 |
- |
4, 5 |
505 |
|
AF026303.1 |
- |
4, 5 |
467 |
|
AF186251.1 |
- |
4, 5 |
700 |
|
AF186252.1 |
- |
4, 5 |
835 |
|
AF186253.1 |
- |
4, 5 |
551 |
|
AF186254.1 |
- |
5, 6 |
733 |
|
AF186255.1 |
- |
5, 6 |
868 |
|
AF186256.1 |
- |
5, 6 |
584 |
|
AK297086.1 |
- |
4, 5 |
641 |
|
AK298306.1 |
- |
4, 5 |
506 |
|
AK313193.1 |
- |
4, 5 |
599 |
|
BC005353.1 |
- |
4, 5 |
452 |
4051264 |
BT006951.1 |
- |
3, 4 |
382 |
|
U66036.1 |
- |
4, 5 |
558 |
|
View Full Product Details