Gene Transcripts
- Gene Symbol
- UBE2D2
- Entrez Gene ID
- 7322
- Gene Name
- ubiquitin conjugating enzyme E2 D2
- Gene Aliases
- E2(17)KB2, PUBC1, UBC4, UBC4/5, UBCH4, UBCH5B
- Chromosome Location
- Chr. 5: 139561166 - 139628434 on Build GRCh38
- UniGene ID
- Hs.108332
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003339.2 |
NP_003330.1
|
2, 3 |
725 |
|
NM_181838.1 |
NP_862821.1
|
3, 4 |
889 |
|
XM_017009820.1 |
XP_016865309.1
|
5, 6 |
1096 |
|
GenBank mRNA |
AF317220.1 |
- |
|
33 |
|
AK001311.1 |
- |
2, 3 |
498 |
|
AK001428.1 |
- |
2, 3 |
552 |
|
AK310273.1 |
- |
2, 3 |
454 |
|
AY651263.1 |
- |
2, 3 |
481 |
|
BC019052.2 |
- |
2, 3 |
509 |
|
BC033349.1 |
- |
2, 3 |
725 |
4827210 |
BQ186099.1 |
- |
3, 4 |
374 |
|
U39317.1 |
- |
2, 3 |
121 |
|
View Full Product Details