Gene Transcripts

Gene Symbol
ZIC1
Entrez Gene ID
7545
Gene Name
Zic family member 1
Gene Aliases
CRS6, ZIC, ZNF201
Chromosome Location
Chr. 3: 147409394 - 147416719 on Build GRCh38
UniGene ID
Hs.598590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003412.3 NP_003403.2 2 1841
GenBank mRNA BC104848.1 - 2 1195
BC104850.1 - 2 1196 8143853
BF920299.1 - 2 138

View Full Product Details