Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304492.1 | NP_001291421.1 | 5 | 532 | |
NM_001304493.1 | NP_001291422.1 | 7 | 657 | ||
NM_001304494.1 | NP_001291423.1 | 3 | 250 | ||
NM_145911.2 | NP_666016.1 | 6 | 1104 | ||
GenBank mRNA | AF507946.1 | - | 6 | 1095 | |
AK091039.1 | - | 5 | 527 | ||
AL080123.1 | - | 1 | 134 | ||
AL833815.1 | - | 2 | 874 | ||
BC007256.1 | - | 6 | 1095 | 3028040 | |
BC048974.1 | - | 7 | 651 | 4816965 | |
BT007400.1 | - | 3 | 281 | ||
BX537507.1 | - | 1 | 2906 |