Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320464.1 | NP_001307393.1 | 6, 7 | 706 | |
NM_024031.3 | NP_076936.1 | 6, 7 | 862 | ||
GenBank mRNA | AK074783.1 | - | 6, 7 | 717 | |
AK125716.1 | - | 4, 5 | 826 | ||
BC000119.1 | - | 6, 7 | 729 | 3351963 | |
BC010232.1 | - | 4, 5 | 471 | ||
BC021934.1 | - | 5, 6 | 814 | 4876482 | |
BC050677.1 | - | 6, 7 | 809 | 6053861 | |
CR936781.1 | - | 4, 5 | 320 |