Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr. 3: 157259742 - 157503626 on Build GRCh38
UniGene ID
Hs.658046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167911.1 NP_001161383.1 12 2328
NM_001167912.1 NP_001161384.1 13 2463
NM_024621.2 NP_078897.2 13 2654
XM_011513134.2 XP_011511436.1 12 2397
XM_011513135.1 XP_011511437.1 13 2293
GenBank mRNA AB051479.1 - 13 2463
AK022666.1 - 3 331
AK027625.1 - 13 2552
AL713656.1 - 12 2251
BC026235.1 - 5 776
BC042159.1 - 6 1113
BC101660.1 - 12 2290 8069166
BC111017.1 - 12 2320 5174889
BC113555.1 - 12 2290 8322607

View Full Product Details