Gene Transcripts

Gene Symbol
MORC4
Entrez Gene ID
79710
Gene Name
MORC family CW-type zinc finger 4
Gene Aliases
ZCW4, ZCWCC2, dJ75H8.2
Chromosome Location
Chr. X: 106940300 - 107000244 on Build GRCh38
UniGene ID
Hs.496544
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085354.2 NP_001078823.1
3, 4 591
NM_024657.4 NP_078933.3
3, 4 591
XM_005262190.3 XP_005262247.1
2, 3 481
XM_017029844.1 XP_016885333.1
2, 3 481
GenBank mRNA AK300305.1 -
2, 3 166
AY927537.1 - LIT3543
AY927537
1, 2 126
CB989767.1 -
3, 4 586
EF125209.1 -
3, 4 316

View Full Product Details