Gene Transcripts
- Gene Symbol
- SLC25A28
- Entrez Gene ID
- 81894
- Gene Name
- solute carrier family 25 member 28
- Gene Aliases
- MFRN2, MRS3/4, MRS4L, NPD016
- Chromosome Location
- Chr. 10: 99610518 - 99659329 on Build GRCh38
- UniGene ID
- Hs.403790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031212.3 |
NP_112489.3
|
3, 4 |
717 |
|
XM_005270208.1 |
XP_005270265.1
|
3, 4 |
707 |
|
XM_005270211.4 |
XP_005270268.1
|
3, 4 |
321 |
|
XM_006718005.3 |
XP_006718068.1
|
3, 4 |
381 |
|
XM_011540239.2 |
XP_011538541.1
|
3, 4 |
603 |
|
XM_011540240.1 |
XP_011538542.1
|
4, 5 |
1853 |
|
XM_017016741.1 |
XP_016872230.1
|
3, 4 |
1789 |
|
XM_017016742.1 |
XP_016872231.1
|
3, 4 |
1610 |
|
GenBank mRNA |
AF267854.1 |
- |
1 |
404 |
|
AF327402.1 |
- |
3, 4 |
617 |
|
AF327403.1 |
- |
3, 4 |
1058 |
|
AJ303077.1 |
- |
3, 4 |
617 |
|
AJ303078.1 |
- |
3, 4 |
1058 |
|
AK056782.1 |
- |
1 |
757 |
|
AL831943.1 |
- |
1 |
280 |
|
BC047312.1 |
- |
3, 4 |
664 |
|
BC058937.1 |
- |
3, 4 |
626 |
6139015 |
BC076399.1 |
- |
3, 4 |
657 |
5728224 |
BC094821.1 |
- |
3, 4 |
400 |
|
View Full Product Details