Gene Transcripts
- Gene Symbol
- WDR54
- Entrez Gene ID
- 84058
- Gene Name
- WD repeat domain 54
- Gene Aliases
- Chromosome Location
- Chr. 2: 74421721 - 74425755 on Build GRCh38
- UniGene ID
- Hs.643480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320823.1 |
NP_001307752.1
|
8, 9 |
860 |
|
NM_001320824.1 |
NP_001307753.1
|
8, 9 |
915 |
|
NM_001320825.1 |
NP_001307754.1
|
7, 8 |
659 |
|
NM_032118.3 |
NP_115494.1
|
8, 9 |
882 |
|
XM_017005064.1 |
XP_016860553.1
|
7, 8 |
661 |
|
GenBank mRNA |
AK023015.1 |
- |
8, 9 |
875 |
|
AK222696.1 |
- |
8, 9 |
875 |
|
AK311621.1 |
- |
3, 4 |
461 |
|
BC051848.1 |
- |
8, 9 |
847 |
6150670 |
CD249537.1 |
- |
7, 8 |
675 |
|
View Full Product Details