Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032806.5 | NP_116195.2 | 2 | 1244 | |
XM_005265515.2 | XP_005265572.1 | 3 | 1375 | ||
XM_011534163.2 | XP_011532465.1 | 3 | 1363 | ||
XM_011534164.1 | XP_011532466.1 | 2 | 1945 | ||
XM_017007353.1 | XP_016862842.1 | 4 | 1454 | ||
GenBank mRNA | AK027472.1 | - | 1 | 504 | |
AK092147.1 | - | 2 | 1032 | ||
AK124737.1 | - | 1 | 2212 | ||
BC060861.1 | - | 2 | 1234 | 30335722 |