Gene Transcripts

Gene Symbol
PGLYRP1
Entrez Gene ID
8993
Gene Name
peptidoglycan recognition protein 1
Gene Aliases
PGLYRP, PGRP, PGRP-S, PGRPS, TAG7, TNFSF3L
Chromosome Location
Chr. 19: 46019153 - 46023298 on Build GRCh38
UniGene ID
Hs.137583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005091.2 NP_005082.1 2 580
GenBank mRNA AF076483.1 - 2 347
AF242517.1 - 2 332
AY358936.1 - 2 328
BC096154.1 - 2 340 40003108
BC096155.3 - 2 340 40003109
BC096156.1 - 2 340 40003112
BC096157.1 - 2 340 40003114
BC101845.1 - 2 342 8069351
BC101847.1 - 2 342 8069353

View Full Product Details