Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321135.1 | NP_001308064.1 | 13 | 1529 | |
NM_001321136.1 | NP_001308065.1 | 15 | 1805 | ||
NM_001321137.1 | NP_001308066.1 | 15 | 1402 | ||
NM_003978.4 | NP_003969.2 | 14 | 1586 | ||
NR_135552.1 | 11 | 1318 | |||
XM_006720737.2 | XP_006720800.1 | 14 | 1318 | ||
XM_011522163.1 | XP_011520465.1 | 15 | 1595 | ||
XM_011522164.1 | XP_011520466.1 | 15 | 1272 | ||
XM_011522165.2 | XP_011520467.1 | 14 | 1127 | ||
XM_011522168.2 | XP_011520470.1 | 15 | 1604 | ||
GenBank mRNA | AB451310.1 | - | 14 | 1097 | |
AB451440.1 | - | 14 | 1097 | ||
AF038602.1 | - | 13 | 1040 | ||
AF038603.1 | - | 14 | 1097 | ||
AK308462.1 | - | 15 | 1436 | ||
AK309856.1 | - | 15 | 1768 | ||
AL110110.1 | - | 3 | 246 | ||
BC008602.1 | - | 14 | 1458 | 4180398 | |
KU178458.1 | - | 14 | 1097 | ||
KU178459.1 | - | 11 | 829 |