Gene Transcripts
- Gene Symbol
- MPDU1
- Entrez Gene ID
- 9526
- Gene Name
- mannose-P-dolichol utilization defect 1
- Gene Aliases
- CDGIF, HBEBP2BPA, Lec35, My008, PP3958, PQLC5, SL15
- Chromosome Location
- Chr. 17: 7583647 - 7588212 on Build GRCh38
- UniGene ID
- Hs.246381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004870.3 |
NP_004861.2
|
6, 7 |
832 |
|
NR_024603.1 |
|
6, 7 |
827 |
|
XM_006721597.2 |
XP_006721660.1
|
5, 6 |
512 |
|
XM_006721598.2 |
XP_006721661.1
|
5, 6 |
774 |
|
XM_006721599.2 |
XP_006721662.1
|
5, 6 |
573 |
|
XM_011524081.2 |
XP_011522383.1
|
6, 7 |
663 |
|
GenBank mRNA |
AF038961.1 |
- |
6, 7 |
632 |
|
AF059752.1 |
- |
6, 7 |
671 |
|
AF258568.1 |
- |
1, 2 |
801 |
|
AK055923.1 |
- |
6, 7 |
547 |
|
AK075299.1 |
- |
6, 7 |
631 |
|
AK225401.1 |
- |
6, 7 |
631 |
|
AK225432.1 |
- |
6, 7 |
627 |
|
AK297001.1 |
- |
5, 6 |
770 |
|
AK297691.1 |
- |
4, 5 |
891 |
|
AK300140.1 |
- |
6, 7 |
618 |
|
AK314527.1 |
- |
6, 7 |
627 |
|
BC001898.1 |
- |
6, 7 |
643 |
3536216 |
BG776573.1 |
- |
2, 3 |
171 |
|
DQ499597.1 |
- |
6, 7 |
614 |
|
KU178478.1 |
- |
6, 7 |
616 |
|
KU178479.1 |
- |
4, 5 |
916 |
|
View Full Product Details