Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282729.1 | NP_001269658.1 | 6 | 1757 | |
NM_001282730.1 | NP_001269659.1 | 6 | 1965 | ||
NM_001282731.1 | NP_001269660.1 | 6 | 1807 | ||
NM_001282732.1 | NP_001269661.1 | 5 | 1347 | ||
NM_014860.2 | NP_055675.1 | 6 | 1752 | ||
XM_005264672.4 | XP_005264729.1 | 6 | 1954 | ||
GenBank mRNA | AB018307.1 | - | 6 | 1494 | |
AF197954.1 | - | 6 | 1513 | ||
AF224759.1 | - | 5 | 2342 | ||
AK000329.1 | - | 6 | 1522 | ||
AK000616.1 | - | 6 | 1308 | ||
AK304889.1 | - | 5 | 1130 | ||
BC074000.1 | - | 6 | 1494 | 6501232 | |
CR456983.1 | - | 4 | 1151 |