Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67508247-67508736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK2AP2
Entrez Gene ID
10263
Gene Name
cyclin dependent kinase 2 associated protein 2
Gene Aliases
DOC-1R, p14
UniGene
Hs.523835
Gene Chromosome Location
Chr.11: 67506490-67508728 on Build GRCh38
Transcripts
NM_005851.4

SNP Detail

SNP ID Chromosome Location
rs1024465296 Chr.11: 67508712-67508712 on Build GRCh38
rs933194834 Chr.11: 67508528-67508528 on Build GRCh38
rs1051597890 Chr.11: 67508540-67508540 on Build GRCh38
rs943281380 Chr.11: 67508566-67508566 on Build GRCh38
rs1871044 Chr.11: 67508687-67508687 on Build GRCh38
rs991924859 Chr.11: 67508423-67508423 on Build GRCh38
rs955701948 Chr.11: 67508426-67508426 on Build GRCh38
rs942858443 Chr.11: 67508571-67508571 on Build GRCh38
rs555674108 Chr.11: 67508558-67508558 on Build GRCh38
rs768451283 Chr.11: 67508379-67508379 on Build GRCh38
rs1036391576 Chr.11: 67508388-67508388 on Build GRCh38
rs777400217 Chr.11: 67508509-67508509 on Build GRCh38
rs1007067952 Chr.11: 67508353-67508353 on Build GRCh38
rs891317659 Chr.11: 67508498-67508498 on Build GRCh38
rs577627008 Chr.11: 67508654-67508654 on Build GRCh38
rs891246127 Chr.11: 67508549-67508549 on Build GRCh38
rs909193759 Chr.11: 67508479-67508479 on Build GRCh38
rs972037947 Chr.11: 67508707-67508707 on Build GRCh38
rs959234061 Chr.11: 67508668-67508668 on Build GRCh38
rs1003073960 Chr.11: 67508648-67508648 on Build GRCh38
rs552808776 Chr.11: 67508565-67508565 on Build GRCh38
rs971602139 Chr.11: 67508491-67508491 on Build GRCh38
rs149084964 Chr.11: 67508651-67508651 on Build GRCh38
rs534582523 Chr.11: 67508536-67508536 on Build GRCh38
rs1051518947 Chr.11: 67508369-67508369 on Build GRCh38
rs933394517 Chr.11: 67508715-67508715 on Build GRCh38
rs968367303 Chr.11: 67508401-67508401 on Build GRCh38
rs544774067 Chr.11: 67508674-67508674 on Build GRCh38
rs988534487 Chr.11: 67508456-67508456 on Build GRCh38
rs765660446 Chr.11: 67508689-67508690 on Build GRCh38
rs539701770 Chr.11: 67508685-67508685 on Build GRCh38
rs1009326093 Chr.11: 67508637-67508637 on Build GRCh38
rs1051144196 Chr.11: 67508308-67508308 on Build GRCh38
rs573582306 Chr.11: 67508530-67508530 on Build GRCh38
rs981690513 Chr.11: 67508335-67508335 on Build GRCh38
rs1053880055 Chr.11: 67508576-67508576 on Build GRCh38
rs918564393 Chr.11: 67508517-67508517 on Build GRCh38
rs575081328 Chr.11: 67508711-67508711 on Build GRCh38
rs893443183 Chr.11: 67508599-67508599 on Build GRCh38
rs751439450 Chr.11: 67508534-67508534 on Build GRCh38
rs1018933336 Chr.11: 67508382-67508382 on Build GRCh38
rs991728592 Chr.11: 67508681-67508681 on Build GRCh38
rs891250387 Chr.11: 67508346-67508346 on Build GRCh38
rs994183428 Chr.11: 67508294-67508294 on Build GRCh38
rs1035847185 Chr.11: 67508653-67508653 on Build GRCh38
rs1051096317 Chr.11: 67508535-67508535 on Build GRCh38
rs927185506 Chr.11: 67508497-67508497 on Build GRCh38
rs987918753 Chr.11: 67508521-67508531 on Build GRCh38
rs958975746 Chr.11: 67508408-67508408 on Build GRCh38
rs942134194 Chr.11: 67508496-67508496 on Build GRCh38
rs370314762 Chr.11: 67508304-67508305 on Build GRCh38
rs906276942 Chr.11: 67508647-67508647 on Build GRCh38
rs190123507 Chr.11: 67508511-67508511 on Build GRCh38
rs1020674836 Chr.11: 67508641-67508641 on Build GRCh38
rs1001087835 Chr.11: 67508677-67508677 on Build GRCh38
rs967503634 Chr.11: 67508311-67508311 on Build GRCh38
rs919095129 Chr.11: 67508520-67508520 on Build GRCh38

View Full Product Details