Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67519755-67520260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP2
Entrez Gene ID
51475
Gene Name
calcium binding protein 2
Gene Aliases
DFNB93
UniGene
Hs.278984
Gene Chromosome Location
Chr.11: 67518947-67523428 on Build GRCh38
Transcripts
NM_001318496.1 NM_016366.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036034 c.532C>T p.R178W Chr.11: 67519898-67519898 on Build GRCh38
4036032 c.618C>T p.D206D Chr.11: 67519812-67519812 on Build GRCh38
5512824 c.637+8delC p.? Chr.11: 67519785-67519785 on Build GRCh38
348527 c.574C>A p.R192R Chr.11: 67519856-67519856 on Build GRCh38
5373384 c.473G>A p.R158Q Chr.11: 67520067-67520067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778190720 Chr.11: 67520058-67520058 on Build GRCh38
rs1045373681 Chr.11: 67519852-67519852 on Build GRCh38
rs753451816 Chr.11: 67519845-67519845 on Build GRCh38
rs185791076 Chr.11: 67520006-67520006 on Build GRCh38
rs758353956 Chr.11: 67519860-67519860 on Build GRCh38
rs771418384 Chr.11: 67520021-67520021 on Build GRCh38
rs1055847679 Chr.11: 67520219-67520219 on Build GRCh38
rs935871525 Chr.11: 67519830-67519830 on Build GRCh38
rs774709291 Chr.11: 67519886-67519886 on Build GRCh38
rs765829174 Chr.11: 67519897-67519897 on Build GRCh38
rs372194472 Chr.11: 67520087-67520087 on Build GRCh38
rs370630219 Chr.11: 67520178-67520178 on Build GRCh38
rs759027635 Chr.11: 67519950-67519950 on Build GRCh38
rs745920263 Chr.11: 67519867-67519867 on Build GRCh38
rs760144485 Chr.11: 67519896-67519896 on Build GRCh38
rs754596713 Chr.11: 67519899-67519899 on Build GRCh38
rs368977197 Chr.11: 67519787-67519787 on Build GRCh38
rs376921045 Chr.11: 67520039-67520039 on Build GRCh38
rs919106845 Chr.11: 67520213-67520213 on Build GRCh38
rs372607625 Chr.11: 67519910-67519910 on Build GRCh38
rs773079612 Chr.11: 67520132-67520132 on Build GRCh38
rs200289196 Chr.11: 67519789-67519789 on Build GRCh38
rs758176096 Chr.11: 67519908-67519908 on Build GRCh38
rs780225393 Chr.11: 67519916-67519916 on Build GRCh38
rs376902077 Chr.11: 67519811-67519811 on Build GRCh38
rs771919679 Chr.11: 67519788-67519788 on Build GRCh38
rs748521430 Chr.11: 67519933-67519933 on Build GRCh38
rs192306176 Chr.11: 67520017-67520017 on Build GRCh38
rs761115561 Chr.11: 67519947-67519947 on Build GRCh38
rs772881310 Chr.11: 67520188-67520188 on Build GRCh38
rs762148757 Chr.11: 67519889-67519889 on Build GRCh38
rs933295107 Chr.11: 67519993-67519993 on Build GRCh38
rs779057963 Chr.11: 67519932-67519932 on Build GRCh38
rs878929048 Chr.11: 67519780-67519780 on Build GRCh38
rs752829239 Chr.11: 67520068-67520068 on Build GRCh38
rs755056946 Chr.11: 67519988-67519988 on Build GRCh38
rs752524599 Chr.11: 67519906-67519906 on Build GRCh38
rs146762535 Chr.11: 67519816-67519816 on Build GRCh38
rs762699596 Chr.11: 67519963-67519963 on Build GRCh38
rs561728040 Chr.11: 67520135-67520135 on Build GRCh38
rs770035101 Chr.11: 67519868-67519868 on Build GRCh38
rs969139121 Chr.11: 67519900-67519901 on Build GRCh38
rs751411709 Chr.11: 67519980-67519980 on Build GRCh38
rs199660389 Chr.11: 67520046-67520046 on Build GRCh38
rs746246468 Chr.11: 67520165-67520165 on Build GRCh38
rs371876888 Chr.11: 67519782-67519782 on Build GRCh38
rs778804226 Chr.11: 67520067-67520067 on Build GRCh38
rs145369252 Chr.11: 67519840-67519840 on Build GRCh38
rs773591861 Chr.11: 67519939-67519939 on Build GRCh38
rs117096937 Chr.11: 67519883-67519883 on Build GRCh38
rs1043567267 Chr.11: 67520048-67520048 on Build GRCh38
rs529226265 Chr.11: 67520221-67520221 on Build GRCh38
rs762321651 Chr.11: 67519823-67519823 on Build GRCh38
rs756574355 Chr.11: 67520167-67520167 on Build GRCh38
rs994382500 Chr.11: 67519994-67519994 on Build GRCh38
rs376707764 Chr.11: 67519786-67519786 on Build GRCh38
rs764868118 Chr.11: 67519905-67519905 on Build GRCh38
rs748780817 Chr.11: 67520175-67520175 on Build GRCh38
rs906919781 Chr.11: 67519872-67519872 on Build GRCh38
rs750365607 Chr.11: 67520060-67520060 on Build GRCh38
rs769148166 Chr.11: 67520036-67520036 on Build GRCh38
rs945878378 Chr.11: 67520083-67520083 on Build GRCh38
rs971919952 Chr.11: 67520186-67520186 on Build GRCh38
rs772297513 Chr.11: 67520007-67520007 on Build GRCh38
rs753361851 Chr.11: 67519898-67519898 on Build GRCh38
rs757194133 Chr.11: 67519981-67519981 on Build GRCh38
rs768484510 Chr.11: 67519779-67519779 on Build GRCh38
rs774793813 Chr.11: 67520096-67520096 on Build GRCh38
rs267603139 Chr.11: 67520115-67520115 on Build GRCh38
rs770128042 Chr.11: 67519798-67519798 on Build GRCh38
rs555794796 Chr.11: 67519875-67519875 on Build GRCh38
rs776117741 Chr.11: 67519948-67519948 on Build GRCh38
rs749928273 Chr.11: 67519833-67519833 on Build GRCh38
rs767911347 Chr.11: 67519828-67519828 on Build GRCh38
rs149712664 Chr.11: 67519792-67519792 on Build GRCh38
rs768052190 Chr.11: 67519892-67519892 on Build GRCh38
rs766408316 Chr.11: 67520137-67520137 on Build GRCh38
rs369999319 Chr.11: 67519819-67519819 on Build GRCh38
rs368278027 Chr.11: 67520084-67520084 on Build GRCh38
rs550353835 Chr.11: 67520201-67520201 on Build GRCh38
rs780668362 Chr.11: 67520172-67520172 on Build GRCh38
rs685350 Chr.11: 67519998-67519998 on Build GRCh38
rs137889404 Chr.11: 67520093-67520093 on Build GRCh38
rs762453427 Chr.11: 67520041-67520041 on Build GRCh38
rs771762987 Chr.11: 67520185-67520185 on Build GRCh38
rs759055794 Chr.11: 67520028-67520028 on Build GRCh38
rs1014475660 Chr.11: 67520094-67520094 on Build GRCh38
rs753936268 Chr.11: 67519986-67519986 on Build GRCh38
rs779161732 Chr.11: 67520001-67520001 on Build GRCh38
rs781344128 Chr.11: 67519982-67519982 on Build GRCh38
rs763377053 Chr.11: 67519814-67519814 on Build GRCh38
rs746902163 Chr.11: 67519911-67519911 on Build GRCh38
rs941741867 Chr.11: 67520232-67520232 on Build GRCh38
rs746312982 Chr.11: 67520091-67520091 on Build GRCh38
rs772359355 Chr.11: 67519935-67519935 on Build GRCh38
rs143579624 Chr.11: 67520074-67520074 on Build GRCh38
rs751566841 Chr.11: 67519917-67519917 on Build GRCh38
rs768317262 Chr.11: 67520176-67520176 on Build GRCh38
rs561090026 Chr.11: 67519841-67519841 on Build GRCh38
rs766208641 Chr.11: 67520211-67520211 on Build GRCh38
rs781355710 Chr.11: 67520163-67520163 on Build GRCh38
rs748874482 Chr.11: 67520102-67520102 on Build GRCh38
rs1025072301 Chr.11: 67519880-67519880 on Build GRCh38
rs371462785 Chr.11: 67520156-67520156 on Build GRCh38
rs747352492 Chr.11: 67520081-67520081 on Build GRCh38
rs761665108 Chr.11: 67520128-67520128 on Build GRCh38
rs553844451 Chr.11: 67519975-67519975 on Build GRCh38
rs904697651 Chr.11: 67519803-67519803 on Build GRCh38
rs368718413 Chr.11: 67519856-67519856 on Build GRCh38
rs980934581 Chr.11: 67519907-67519907 on Build GRCh38
rs199708887 Chr.11: 67519844-67519844 on Build GRCh38
rs369233041 Chr.11: 67520140-67520140 on Build GRCh38
rs764921458 Chr.11: 67519955-67519955 on Build GRCh38
rs768914726 Chr.11: 67519923-67519923 on Build GRCh38
rs114668091 Chr.11: 67519920-67519920 on Build GRCh38
rs760580604 Chr.11: 67520206-67520206 on Build GRCh38
rs777631985 Chr.11: 67519861-67519861 on Build GRCh38
rs754614044 Chr.11: 67519855-67519855 on Build GRCh38
rs768409215 Chr.11: 67520106-67520106 on Build GRCh38
rs767378517 Chr.11: 67520130-67520130 on Build GRCh38
rs757281872 Chr.11: 67519914-67519914 on Build GRCh38
rs773753281 Chr.11: 67519893-67519893 on Build GRCh38
rs751975494 Chr.11: 67520163-67520163 on Build GRCh38
rs368437370 Chr.11: 67520059-67520059 on Build GRCh38
rs969469611 Chr.11: 67520123-67520123 on Build GRCh38
rs778047665 Chr.11: 67520012-67520012 on Build GRCh38
rs758907938 Chr.11: 67520020-67520020 on Build GRCh38
rs1017085785 Chr.11: 67519926-67519926 on Build GRCh38
rs764270235 Chr.11: 67520146-67520146 on Build GRCh38
rs866283688 Chr.11: 67520095-67520095 on Build GRCh38
rs774192693 Chr.11: 67520118-67520118 on Build GRCh38
rs954587420 Chr.11: 67520216-67520216 on Build GRCh38
rs543555075 Chr.11: 67520057-67520057 on Build GRCh38
rs752334231 Chr.11: 67519962-67519962 on Build GRCh38
rs183733161 Chr.11: 67520239-67520239 on Build GRCh38
rs747750141 Chr.11: 67520179-67520179 on Build GRCh38
rs761754845 Chr.11: 67520056-67520056 on Build GRCh38
rs756105648 Chr.11: 67520062-67520062 on Build GRCh38

View Full Product Details