Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67521845-67522353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP2
Entrez Gene ID
51475
Gene Name
calcium binding protein 2
Gene Aliases
DFNB93
UniGene
Hs.278984
Gene Chromosome Location
Chr.11: 67518947-67523428 on Build GRCh38
Transcripts
NM_001318496.1 NM_016366.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4799193 c.240T>C p.I80I Chr.11: 67521956-67521956 on Build GRCh38
5794710 c.244+7G>T p.? Chr.11: 67521945-67521945 on Build GRCh38
4816864 c.232G>A p.E78K Chr.11: 67521964-67521964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540712728 Chr.11: 67522306-67522306 on Build GRCh38
rs771258675 Chr.11: 67521952-67521952 on Build GRCh38
rs1054195353 Chr.11: 67521942-67521942 on Build GRCh38
rs556941177 Chr.11: 67522112-67522112 on Build GRCh38
rs532726960 Chr.11: 67522318-67522318 on Build GRCh38
rs1047568860 Chr.11: 67522058-67522058 on Build GRCh38
rs577400882 Chr.11: 67522090-67522090 on Build GRCh38
rs763287401 Chr.11: 67522011-67522011 on Build GRCh38
rs779763116 Chr.11: 67521979-67521979 on Build GRCh38
rs112972069 Chr.11: 67522203-67522203 on Build GRCh38
rs977542222 Chr.11: 67521943-67521943 on Build GRCh38
rs765901779 Chr.11: 67521970-67521970 on Build GRCh38
rs937274105 Chr.11: 67522167-67522167 on Build GRCh38
rs732288 Chr.11: 67521915-67521915 on Build GRCh38
rs890378868 Chr.11: 67522066-67522066 on Build GRCh38
rs573413271 Chr.11: 67522248-67522248 on Build GRCh38
rs561328889 Chr.11: 67522204-67522204 on Build GRCh38
rs769182345 Chr.11: 67522012-67522012 on Build GRCh38
rs957069507 Chr.11: 67522328-67522328 on Build GRCh38
rs779851068 Chr.11: 67521913-67521913 on Build GRCh38
rs899228112 Chr.11: 67522020-67522020 on Build GRCh38
rs538576070 Chr.11: 67522091-67522091 on Build GRCh38
rs200976551 Chr.11: 67521944-67521944 on Build GRCh38
rs924879386 Chr.11: 67521957-67521957 on Build GRCh38
rs553244596 Chr.11: 67522123-67522123 on Build GRCh38
rs766960942 Chr.11: 67522029-67522029 on Build GRCh38
rs150664916 Chr.11: 67521969-67521969 on Build GRCh38
rs575603514 Chr.11: 67522117-67522117 on Build GRCh38
rs150573159 Chr.11: 67521990-67521990 on Build GRCh38
rs532889037 Chr.11: 67521911-67521911 on Build GRCh38
rs944530979 Chr.11: 67522297-67522297 on Build GRCh38
rs1047600404 Chr.11: 67522247-67522247 on Build GRCh38
rs550969210 Chr.11: 67521935-67521935 on Build GRCh38
rs765647878 Chr.11: 67521965-67521965 on Build GRCh38
rs768487955 Chr.11: 67521934-67521934 on Build GRCh38
rs767124487 Chr.11: 67521976-67521976 on Build GRCh38
rs143510168 Chr.11: 67522084-67522084 on Build GRCh38
rs376333735 Chr.11: 67521972-67521972 on Build GRCh38
rs373295487 Chr.11: 67522051-67522051 on Build GRCh38
rs993126587 Chr.11: 67522152-67522152 on Build GRCh38
rs995591399 Chr.11: 67522035-67522035 on Build GRCh38
rs565467704 Chr.11: 67521873-67521873 on Build GRCh38
rs367669139 Chr.11: 67521866-67521866 on Build GRCh38
rs935713157 Chr.11: 67522118-67522118 on Build GRCh38
rs762338446 Chr.11: 67522023-67522023 on Build GRCh38
rs916132735 Chr.11: 67522159-67522159 on Build GRCh38
rs960482031 Chr.11: 67522137-67522137 on Build GRCh38
rs1005009026 Chr.11: 67521892-67521892 on Build GRCh38
rs970105933 Chr.11: 67522200-67522200 on Build GRCh38
rs373352152 Chr.11: 67522014-67522014 on Build GRCh38
rs114359066 Chr.11: 67522074-67522074 on Build GRCh38
rs964306449 Chr.11: 67522282-67522282 on Build GRCh38
rs929343280 Chr.11: 67522242-67522242 on Build GRCh38
rs369309124 Chr.11: 67522004-67522004 on Build GRCh38
rs139560872 Chr.11: 67522083-67522083 on Build GRCh38
rs770146508 Chr.11: 67522005-67522005 on Build GRCh38
rs951557794 Chr.11: 67522056-67522056 on Build GRCh38
rs1008426711 Chr.11: 67522241-67522241 on Build GRCh38
rs759873671 Chr.11: 67521958-67521958 on Build GRCh38
rs935768635 Chr.11: 67521928-67521928 on Build GRCh38
rs945924140 Chr.11: 67522149-67522149 on Build GRCh38
rs921733266 Chr.11: 67521875-67521875 on Build GRCh38
rs749039443 Chr.11: 67521989-67521989 on Build GRCh38
rs34486649 Chr.11: 67522095-67522096 on Build GRCh38
rs549074072 Chr.11: 67522070-67522070 on Build GRCh38
rs776877052 Chr.11: 67521999-67521999 on Build GRCh38
rs749944898 Chr.11: 67522030-67522030 on Build GRCh38
rs750036879 Chr.11: 67521978-67521978 on Build GRCh38
rs747083299 Chr.11: 67521997-67521997 on Build GRCh38
rs886430025 Chr.11: 67521878-67521878 on Build GRCh38
rs1055759212 Chr.11: 67522168-67522168 on Build GRCh38
rs1028582021 Chr.11: 67522320-67522321 on Build GRCh38
rs995580265 Chr.11: 67522178-67522178 on Build GRCh38
rs749196636 Chr.11: 67521916-67521916 on Build GRCh38
rs1022024890 Chr.11: 67522094-67522094 on Build GRCh38
rs775919990 Chr.11: 67522010-67522010 on Build GRCh38
rs1038979789 Chr.11: 67522299-67522299 on Build GRCh38
rs907089151 Chr.11: 67522160-67522160 on Build GRCh38
rs774758977 Chr.11: 67522018-67522018 on Build GRCh38
rs565505646 Chr.11: 67522307-67522307 on Build GRCh38
rs771098980 Chr.11: 67521998-67521998 on Build GRCh38
rs998985810 Chr.11: 67522116-67522116 on Build GRCh38
rs777036556 Chr.11: 67521955-67521955 on Build GRCh38

View Full Product Details