Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67642463-67642967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACY3
Entrez Gene ID
91703
Gene Name
aminoacylase 3
Gene Aliases
ACY-3, HCBP1
UniGene
Hs.126265
Gene Chromosome Location
Chr.11: 67642551-67650829 on Build GRCh38
Transcripts
NM_080658.1 XM_017018549.1 XM_017018550.1 XM_017018551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2042299 c.913G>A p.V305M Chr.11: 67642771-67642771 on Build GRCh38
6519294 c.870G>T p.E290D Chr.11: 67642814-67642814 on Build GRCh38
2042301 c.853G>A p.E285K Chr.11: 67642831-67642831 on Build GRCh38
2042297 c.930G>A p.A310A Chr.11: 67642754-67642754 on Build GRCh38
2042298 c.928G>A p.A310T Chr.11: 67642756-67642756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577202816 Chr.11: 67642527-67642527 on Build GRCh38
rs149837610 Chr.11: 67642757-67642757 on Build GRCh38
rs139551384 Chr.11: 67642739-67642739 on Build GRCh38
rs202084142 Chr.11: 67642831-67642831 on Build GRCh38
rs758309501 Chr.11: 67642547-67642547 on Build GRCh38
rs2290959 Chr.11: 67642843-67642843 on Build GRCh38
rs764559224 Chr.11: 67642836-67642836 on Build GRCh38
rs774405351 Chr.11: 67642737-67642737 on Build GRCh38
rs1032914870 Chr.11: 67642551-67642551 on Build GRCh38
rs146829262 Chr.11: 67642854-67642854 on Build GRCh38
rs756917923 Chr.11: 67642672-67642672 on Build GRCh38
rs140495037 Chr.11: 67642935-67642935 on Build GRCh38
rs776815218 Chr.11: 67642808-67642808 on Build GRCh38
rs758845029 Chr.11: 67642747-67642747 on Build GRCh38
rs762743990 Chr.11: 67642722-67642722 on Build GRCh38
rs546205632 Chr.11: 67642645-67642645 on Build GRCh38
rs752171276 Chr.11: 67642754-67642754 on Build GRCh38
rs763284642 Chr.11: 67642944-67642944 on Build GRCh38
rs947767665 Chr.11: 67642587-67642587 on Build GRCh38
rs779695753 Chr.11: 67642761-67642761 on Build GRCh38
rs966298159 Chr.11: 67642515-67642515 on Build GRCh38
rs958440108 Chr.11: 67642627-67642627 on Build GRCh38
rs1019008656 Chr.11: 67642499-67642499 on Build GRCh38
rs1005413637 Chr.11: 67642496-67642496 on Build GRCh38
rs115044142 Chr.11: 67642555-67642555 on Build GRCh38
rs146627401 Chr.11: 67642832-67642832 on Build GRCh38
rs202083681 Chr.11: 67642755-67642755 on Build GRCh38
rs775215471 Chr.11: 67642715-67642715 on Build GRCh38
rs774128346 Chr.11: 67642780-67642780 on Build GRCh38
rs980456062 Chr.11: 67642803-67642803 on Build GRCh38
rs367629312 Chr.11: 67642675-67642675 on Build GRCh38
rs1010193761 Chr.11: 67642560-67642560 on Build GRCh38
rs143710293 Chr.11: 67642853-67642853 on Build GRCh38
rs1035349043 Chr.11: 67642798-67642798 on Build GRCh38
rs748170053 Chr.11: 67642793-67642793 on Build GRCh38
rs572426815 Chr.11: 67642640-67642640 on Build GRCh38
rs569162389 Chr.11: 67642707-67642707 on Build GRCh38
rs775632168 Chr.11: 67642734-67642735 on Build GRCh38
rs574543962 Chr.11: 67642602-67642602 on Build GRCh38
rs778125820 Chr.11: 67642750-67642750 on Build GRCh38
rs368425963 Chr.11: 67642678-67642678 on Build GRCh38
rs145815101 Chr.11: 67642772-67642772 on Build GRCh38
rs371660515 Chr.11: 67642877-67642877 on Build GRCh38
rs539368779 Chr.11: 67642844-67642844 on Build GRCh38
rs185370494 Chr.11: 67642840-67642840 on Build GRCh38
rs754159939 Chr.11: 67642673-67642673 on Build GRCh38
rs780704607 Chr.11: 67642704-67642704 on Build GRCh38
rs947833560 Chr.11: 67642779-67642779 on Build GRCh38
rs768420616 Chr.11: 67642723-67642723 on Build GRCh38
rs368141215 Chr.11: 67642822-67642822 on Build GRCh38
rs780801563 Chr.11: 67642756-67642756 on Build GRCh38
rs773292003 Chr.11: 67642879-67642879 on Build GRCh38
rs974615380 Chr.11: 67642522-67642522 on Build GRCh38
rs765589639 Chr.11: 67642741-67642741 on Build GRCh38
rs775820522 Chr.11: 67642936-67642936 on Build GRCh38
rs556429575 Chr.11: 67642570-67642570 on Build GRCh38
rs1044562617 Chr.11: 67642596-67642596 on Build GRCh38
rs145716789 Chr.11: 67642609-67642609 on Build GRCh38
rs368510323 Chr.11: 67642771-67642771 on Build GRCh38
rs78344373 Chr.11: 67642929-67642929 on Build GRCh38
rs770172443 Chr.11: 67642711-67642711 on Build GRCh38
rs371929650 Chr.11: 67642785-67642785 on Build GRCh38
rs181300164 Chr.11: 67642661-67642661 on Build GRCh38
rs778344985 Chr.11: 67642693-67642693 on Build GRCh38
rs770528217 Chr.11: 67642696-67642696 on Build GRCh38
rs199948169 Chr.11: 67642683-67642683 on Build GRCh38
rs2290960 Chr.11: 67642603-67642603 on Build GRCh38
rs771927993 Chr.11: 67642807-67642807 on Build GRCh38
rs370053612 Chr.11: 67642714-67642714 on Build GRCh38
rs368895526 Chr.11: 67642572-67642574 on Build GRCh38
rs976727601 Chr.11: 67642878-67642878 on Build GRCh38
rs930906173 Chr.11: 67642608-67642608 on Build GRCh38
rs188330165 Chr.11: 67642868-67642868 on Build GRCh38
rs901806839 Chr.11: 67642862-67642862 on Build GRCh38
rs570722268 Chr.11: 67642847-67642847 on Build GRCh38
rs749879139 Chr.11: 67642846-67642846 on Build GRCh38
rs142983989 Chr.11: 67642824-67642824 on Build GRCh38
rs868390081 Chr.11: 67642790-67642791 on Build GRCh38
rs112020666 Chr.11: 67642658-67642658 on Build GRCh38
rs779677196 Chr.11: 67642852-67642852 on Build GRCh38
rs753989539 Chr.11: 67642740-67642740 on Build GRCh38
rs765614655 Chr.11: 67642856-67642856 on Build GRCh38
rs928390109 Chr.11: 67642622-67642622 on Build GRCh38
rs866865385 Chr.11: 67642524-67642524 on Build GRCh38
rs367771910 Chr.11: 67642573-67642575 on Build GRCh38
rs923635127 Chr.11: 67642549-67642549 on Build GRCh38
rs763229827 Chr.11: 67642844-67642844 on Build GRCh38
rs758931906 Chr.11: 67642691-67642691 on Build GRCh38
rs550546154 Chr.11: 67642744-67642744 on Build GRCh38
rs535527775 Chr.11: 67642928-67642928 on Build GRCh38
rs148549665 Chr.11: 67642818-67642818 on Build GRCh38
rs917661675 Chr.11: 67642583-67642583 on Build GRCh38
rs745558912 Chr.11: 67642710-67642710 on Build GRCh38

View Full Product Details