Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67642712-67643154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACY3
Entrez Gene ID
91703
Gene Name
aminoacylase 3
Gene Aliases
ACY-3, HCBP1
UniGene
Hs.126265
Gene Chromosome Location
Chr.11: 67642551-67650829 on Build GRCh38
Transcripts
NM_080658.1 XM_017018549.1 XM_017018550.1 XM_017018551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2042299 c.913G>A p.V305M Chr.11: 67642771-67642771 on Build GRCh38
6519294 c.870G>T p.E290D Chr.11: 67642814-67642814 on Build GRCh38
2042301 c.853G>A p.E285K Chr.11: 67642831-67642831 on Build GRCh38
2042297 c.930G>A p.A310A Chr.11: 67642754-67642754 on Build GRCh38
2042298 c.928G>A p.A310T Chr.11: 67642756-67642756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149837610 Chr.11: 67642757-67642757 on Build GRCh38
rs139551384 Chr.11: 67642739-67642739 on Build GRCh38
rs202084142 Chr.11: 67642831-67642831 on Build GRCh38
rs2290959 Chr.11: 67642843-67642843 on Build GRCh38
rs774405351 Chr.11: 67642737-67642737 on Build GRCh38
rs764559224 Chr.11: 67642836-67642836 on Build GRCh38
rs180803100 Chr.11: 67642947-67642947 on Build GRCh38
rs146829262 Chr.11: 67642854-67642854 on Build GRCh38
rs140495037 Chr.11: 67642935-67642935 on Build GRCh38
rs758845029 Chr.11: 67642747-67642747 on Build GRCh38
rs776815218 Chr.11: 67642808-67642808 on Build GRCh38
rs752171276 Chr.11: 67642754-67642754 on Build GRCh38
rs919661148 Chr.11: 67643036-67643036 on Build GRCh38
rs763284642 Chr.11: 67642944-67642944 on Build GRCh38
rs755591200 Chr.11: 67642967-67642967 on Build GRCh38
rs779695753 Chr.11: 67642761-67642761 on Build GRCh38
rs952478210 Chr.11: 67643110-67643110 on Build GRCh38
rs146627401 Chr.11: 67642832-67642832 on Build GRCh38
rs750996158 Chr.11: 67642959-67642959 on Build GRCh38
rs202083681 Chr.11: 67642755-67642755 on Build GRCh38
rs774128346 Chr.11: 67642780-67642780 on Build GRCh38
rs980456062 Chr.11: 67642803-67642803 on Build GRCh38
rs969020719 Chr.11: 67643001-67643001 on Build GRCh38
rs771059340 Chr.11: 67643090-67643090 on Build GRCh38
rs1035349043 Chr.11: 67642798-67642798 on Build GRCh38
rs143710293 Chr.11: 67642853-67642853 on Build GRCh38
rs748170053 Chr.11: 67642793-67642793 on Build GRCh38
rs747916053 Chr.11: 67642986-67642986 on Build GRCh38
rs187434412 Chr.11: 67642983-67642983 on Build GRCh38
rs775632168 Chr.11: 67642734-67642735 on Build GRCh38
rs562441457 Chr.11: 67643100-67643100 on Build GRCh38
rs778125820 Chr.11: 67642750-67642750 on Build GRCh38
rs374625311 Chr.11: 67642971-67642971 on Build GRCh38
rs768001633 Chr.11: 67642957-67642957 on Build GRCh38
rs371660515 Chr.11: 67642877-67642877 on Build GRCh38
rs145815101 Chr.11: 67642772-67642772 on Build GRCh38
rs539368779 Chr.11: 67642844-67642844 on Build GRCh38
rs185370494 Chr.11: 67642840-67642840 on Build GRCh38
rs947833560 Chr.11: 67642779-67642779 on Build GRCh38
rs906816531 Chr.11: 67643000-67643000 on Build GRCh38
rs368141215 Chr.11: 67642822-67642822 on Build GRCh38
rs773292003 Chr.11: 67642879-67642879 on Build GRCh38
rs780801563 Chr.11: 67642756-67642756 on Build GRCh38
rs774736010 Chr.11: 67642950-67642950 on Build GRCh38
rs765589639 Chr.11: 67642741-67642741 on Build GRCh38
rs775820522 Chr.11: 67642936-67642936 on Build GRCh38
rs1031223946 Chr.11: 67642969-67642969 on Build GRCh38
rs148125347 Chr.11: 67643069-67643069 on Build GRCh38
rs373512387 Chr.11: 67642976-67642976 on Build GRCh38
rs368510323 Chr.11: 67642771-67642771 on Build GRCh38
rs1012681227 Chr.11: 67643085-67643085 on Build GRCh38
rs78344373 Chr.11: 67642929-67642929 on Build GRCh38
rs371929650 Chr.11: 67642785-67642785 on Build GRCh38
rs771927993 Chr.11: 67642807-67642807 on Build GRCh38
rs976727601 Chr.11: 67642878-67642878 on Build GRCh38
rs901806839 Chr.11: 67642862-67642862 on Build GRCh38
rs188330165 Chr.11: 67642868-67642868 on Build GRCh38
rs570722268 Chr.11: 67642847-67642847 on Build GRCh38
rs980531147 Chr.11: 67643032-67643032 on Build GRCh38
rs749879139 Chr.11: 67642846-67642846 on Build GRCh38
rs142983989 Chr.11: 67642824-67642824 on Build GRCh38
rs868390081 Chr.11: 67642790-67642791 on Build GRCh38
rs1024652910 Chr.11: 67642997-67642997 on Build GRCh38
rs779677196 Chr.11: 67642852-67642852 on Build GRCh38
rs753989539 Chr.11: 67642740-67642740 on Build GRCh38
rs576628970 Chr.11: 67643015-67643015 on Build GRCh38
rs762099774 Chr.11: 67642954-67642954 on Build GRCh38
rs765614655 Chr.11: 67642856-67642856 on Build GRCh38
rs763229827 Chr.11: 67642844-67642844 on Build GRCh38
rs550546154 Chr.11: 67642744-67642744 on Build GRCh38
rs535527775 Chr.11: 67642928-67642928 on Build GRCh38
rs907995082 Chr.11: 67643103-67643103 on Build GRCh38
rs200337718 Chr.11: 67642952-67642952 on Build GRCh38
rs148549665 Chr.11: 67642818-67642818 on Build GRCh38

View Full Product Details