Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68008494-68008989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
UniGene
Hs.523841
Gene Chromosome Location
Chr.11: 68008547-68029282 on Build GRCh38
Transcripts
NM_001161473.2

SNP Detail

SNP ID Chromosome Location
rs1049455457 Chr.11: 68008714-68008714 on Build GRCh38
rs572263877 Chr.11: 68008709-68008709 on Build GRCh38
rs371095527 Chr.11: 68008721-68008722 on Build GRCh38
rs933546587 Chr.11: 68008718-68008718 on Build GRCh38
rs308337 Chr.11: 68008822-68008822 on Build GRCh38
rs542479972 Chr.11: 68008653-68008653 on Build GRCh38
rs548725752 Chr.11: 68008714-68008714 on Build GRCh38
rs909758802 Chr.11: 68008654-68008654 on Build GRCh38
rs866858292 Chr.11: 68008878-68008878 on Build GRCh38
rs564635081 Chr.11: 68008766-68008766 on Build GRCh38
rs1055306252 Chr.11: 68008943-68008943 on Build GRCh38
rs878994994 Chr.11: 68008617-68008617 on Build GRCh38
rs1033794746 Chr.11: 68008645-68008645 on Build GRCh38
rs903518106 Chr.11: 68008561-68008561 on Build GRCh38
rs182488365 Chr.11: 68008601-68008601 on Build GRCh38
rs935631319 Chr.11: 68008719-68008719 on Build GRCh38
rs184759902 Chr.11: 68008883-68008883 on Build GRCh38
rs1002269427 Chr.11: 68008849-68008849 on Build GRCh38
rs988051543 Chr.11: 68008722-68008722 on Build GRCh38
rs951587652 Chr.11: 68008776-68008776 on Build GRCh38
rs193115937 Chr.11: 68008860-68008860 on Build GRCh38
rs959330252 Chr.11: 68008646-68008646 on Build GRCh38
rs1017338593 Chr.11: 68008819-68008819 on Build GRCh38
rs140148305 Chr.11: 68008917-68008917 on Build GRCh38
rs368227124 Chr.11: 68008817-68008818 on Build GRCh38
rs984102976 Chr.11: 68008814-68008814 on Build GRCh38
rs758119464 Chr.11: 68008765-68008765 on Build GRCh38
rs971881260 Chr.11: 68008713-68008713 on Build GRCh38
rs752012516 Chr.11: 68008841-68008841 on Build GRCh38
rs376286126 Chr.11: 68008683-68008683 on Build GRCh38
rs187760286 Chr.11: 68008710-68008710 on Build GRCh38
rs972709849 Chr.11: 68008682-68008682 on Build GRCh38
rs308338 Chr.11: 68008772-68008772 on Build GRCh38
rs1003672875 Chr.11: 68008939-68008939 on Build GRCh38
rs1039420418 Chr.11: 68008560-68008560 on Build GRCh38
rs544917080 Chr.11: 68008630-68008633 on Build GRCh38
rs879805030 Chr.11: 68008777-68008777 on Build GRCh38
rs973042936 Chr.11: 68008834-68008834 on Build GRCh38
rs970927415 Chr.11: 68008605-68008605 on Build GRCh38
rs117602660 Chr.11: 68008813-68008813 on Build GRCh38
rs189253486 Chr.11: 68008887-68008887 on Build GRCh38
rs574754700 Chr.11: 68008628-68008628 on Build GRCh38
rs748529059 Chr.11: 68008918-68008918 on Build GRCh38
rs1009704216 Chr.11: 68008589-68008589 on Build GRCh38
rs955179071 Chr.11: 68008847-68008847 on Build GRCh38
rs3763941 Chr.11: 68008892-68008892 on Build GRCh38
rs939938237 Chr.11: 68008667-68008667 on Build GRCh38
rs571719186 Chr.11: 68008944-68008944 on Build GRCh38
rs963404346 Chr.11: 68008877-68008877 on Build GRCh38
rs891495014 Chr.11: 68008753-68008753 on Build GRCh38
rs308336 Chr.11: 68008850-68008850 on Build GRCh38
rs1003728567 Chr.11: 68008806-68008806 on Build GRCh38
rs1054538088 Chr.11: 68008749-68008749 on Build GRCh38
rs889551429 Chr.11: 68008716-68008716 on Build GRCh38
rs550420410 Chr.11: 68008809-68008809 on Build GRCh38
rs910938077 Chr.11: 68008871-68008871 on Build GRCh38
rs5792428 Chr.11: 68008711-68008712 on Build GRCh38
rs148681395 Chr.11: 68008576-68008577 on Build GRCh38
rs945761953 Chr.11: 68008907-68008907 on Build GRCh38
rs772445586 Chr.11: 68008936-68008936 on Build GRCh38

View Full Product Details