Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68019589-68020093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
UniGene
Hs.523841
Gene Chromosome Location
Chr.11: 68008547-68029282 on Build GRCh38
Transcripts
NM_000694.3 NM_001030010.2 NM_001161473.2 NM_001290058.1 NM_001290059.1

SNP Detail

SNP ID Chromosome Location
rs397843672 Chr.11: 68019912-68019913 on Build GRCh38
rs987961490 Chr.11: 68019854-68019854 on Build GRCh38
rs781729019 Chr.11: 68019690-68019690 on Build GRCh38
rs61887541 Chr.11: 68019671-68019671 on Build GRCh38
rs1023725096 Chr.11: 68019898-68019898 on Build GRCh38
rs780763824 Chr.11: 68019900-68019901 on Build GRCh38
rs181770776 Chr.11: 68019956-68019956 on Build GRCh38
rs201905211 Chr.11: 68019739-68019739 on Build GRCh38
rs570595291 Chr.11: 68019733-68019733 on Build GRCh38
rs955788646 Chr.11: 68019831-68019831 on Build GRCh38
rs774202315 Chr.11: 68019738-68019738 on Build GRCh38
rs942512453 Chr.11: 68020028-68020028 on Build GRCh38
rs535721373 Chr.11: 68019650-68019650 on Build GRCh38
rs369340202 Chr.11: 68019753-68019753 on Build GRCh38
rs548073091 Chr.11: 68019663-68019663 on Build GRCh38
rs960875843 Chr.11: 68019860-68019860 on Build GRCh38
rs773258034 Chr.11: 68019748-68019748 on Build GRCh38
rs769378528 Chr.11: 68019706-68019706 on Build GRCh38
rs982146951 Chr.11: 68019897-68019898 on Build GRCh38
rs375894932 Chr.11: 68019768-68019768 on Build GRCh38
rs999757237 Chr.11: 68019817-68019817 on Build GRCh38
rs561898756 Chr.11: 68019632-68019632 on Build GRCh38
rs777455902 Chr.11: 68019758-68019758 on Build GRCh38
rs975533120 Chr.11: 68020051-68020051 on Build GRCh38
rs769273922 Chr.11: 68019774-68019774 on Build GRCh38
rs772596233 Chr.11: 68019898-68019901 on Build GRCh38
rs554049096 Chr.11: 68019804-68019804 on Build GRCh38
rs750375695 Chr.11: 68019747-68019747 on Build GRCh38
rs1037960631 Chr.11: 68019981-68019981 on Build GRCh38
rs963151975 Chr.11: 68019929-68019929 on Build GRCh38
rs199800006 Chr.11: 68019778-68019778 on Build GRCh38
rs761445732 Chr.11: 68019797-68019797 on Build GRCh38
rs937960556 Chr.11: 68019925-68019925 on Build GRCh38
rs918501043 Chr.11: 68019752-68019752 on Build GRCh38
rs368403076 Chr.11: 68019810-68019810 on Build GRCh38
rs999641051 Chr.11: 68019660-68019660 on Build GRCh38
rs778270013 Chr.11: 68019759-68019759 on Build GRCh38
rs757647123 Chr.11: 68019766-68019766 on Build GRCh38
rs962762491 Chr.11: 68019727-68019727 on Build GRCh38
rs758732946 Chr.11: 68019754-68019754 on Build GRCh38
rs886899609 Chr.11: 68020057-68020057 on Build GRCh38
rs77192555 Chr.11: 68019670-68019670 on Build GRCh38
rs746412171 Chr.11: 68019771-68019771 on Build GRCh38
rs764346048 Chr.11: 68019830-68019830 on Build GRCh38
rs760714030 Chr.11: 68019805-68019805 on Build GRCh38
rs763140110 Chr.11: 68019816-68019816 on Build GRCh38
rs914319989 Chr.11: 68020052-68020052 on Build GRCh38
rs775078460 Chr.11: 68019716-68019716 on Build GRCh38
rs781306604 Chr.11: 68019832-68019832 on Build GRCh38
rs951261738 Chr.11: 68019973-68019973 on Build GRCh38
rs1019291273 Chr.11: 68020064-68020064 on Build GRCh38
rs771930196 Chr.11: 68019800-68019800 on Build GRCh38
rs537955546 Chr.11: 68019783-68019783 on Build GRCh38
rs2286169 Chr.11: 68020059-68020059 on Build GRCh38
rs190144520 Chr.11: 68019705-68019705 on Build GRCh38
rs554078182 Chr.11: 68019995-68019995 on Build GRCh38
rs771829047 Chr.11: 68019753-68019753 on Build GRCh38
rs1039490280 Chr.11: 68019801-68019801 on Build GRCh38
rs757517031 Chr.11: 68019833-68019833 on Build GRCh38
rs762645901 Chr.11: 68019725-68019725 on Build GRCh38
rs974096154 Chr.11: 68019940-68019940 on Build GRCh38
rs372453336 Chr.11: 68019809-68019809 on Build GRCh38
rs572146256 Chr.11: 68020006-68020006 on Build GRCh38
rs986728080 Chr.11: 68020005-68020005 on Build GRCh38
rs748554044 Chr.11: 68019872-68019872 on Build GRCh38
rs767295715 Chr.11: 68019746-68019746 on Build GRCh38
rs751814182 Chr.11: 68019765-68019765 on Build GRCh38
rs4646811 Chr.11: 68019898-68019899 on Build GRCh38
rs748938903 Chr.11: 68019777-68019777 on Build GRCh38
rs958127355 Chr.11: 68019836-68019836 on Build GRCh38
rs372778000 Chr.11: 68019763-68019763 on Build GRCh38
rs568130666 Chr.11: 68019613-68019613 on Build GRCh38
rs546010003 Chr.11: 68020048-68020048 on Build GRCh38

View Full Product Details