Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68027542-68028051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
UniGene
Hs.523841
Gene Chromosome Location
Chr.11: 68008547-68029282 on Build GRCh38
Transcripts
NM_000694.3 NM_001030010.2 NM_001161473.2 NM_001290058.1 NM_001290059.1

SNP Detail

SNP ID Chromosome Location
rs780683696 Chr.11: 68028005-68028005 on Build GRCh38
rs780058209 Chr.11: 68027870-68027870 on Build GRCh38
rs1056246896 Chr.11: 68027609-68027609 on Build GRCh38
rs541303778 Chr.11: 68028017-68028017 on Build GRCh38
rs796560803 Chr.11: 68027577-68027577 on Build GRCh38
rs748977618 Chr.11: 68027638-68027638 on Build GRCh38
rs373528981 Chr.11: 68027864-68027864 on Build GRCh38
rs1006099560 Chr.11: 68027678-68027678 on Build GRCh38
rs908545113 Chr.11: 68027562-68027562 on Build GRCh38
rs111444623 Chr.11: 68028022-68028022 on Build GRCh38
rs766653976 Chr.11: 68027812-68027812 on Build GRCh38
rs371253831 Chr.11: 68027767-68027767 on Build GRCh38
rs563316427 Chr.11: 68027661-68027661 on Build GRCh38
rs781479683 Chr.11: 68027988-68027988 on Build GRCh38
rs748360551 Chr.11: 68027766-68027766 on Build GRCh38
rs761396208 Chr.11: 68028020-68028020 on Build GRCh38
rs932530075 Chr.11: 68028001-68028001 on Build GRCh38
rs908509521 Chr.11: 68027811-68027811 on Build GRCh38
rs530669407 Chr.11: 68027663-68027663 on Build GRCh38
rs768274330 Chr.11: 68028014-68028014 on Build GRCh38
rs1014367967 Chr.11: 68027701-68027701 on Build GRCh38
rs558740178 Chr.11: 68027930-68027930 on Build GRCh38
rs308341 Chr.11: 68027831-68027831 on Build GRCh38
rs1044704549 Chr.11: 68027640-68027640 on Build GRCh38
rs368554544 Chr.11: 68027808-68027808 on Build GRCh38
rs368275772 Chr.11: 68027877-68027877 on Build GRCh38
rs943928590 Chr.11: 68027563-68027563 on Build GRCh38
rs200709828 Chr.11: 68027914-68027914 on Build GRCh38
rs778759508 Chr.11: 68027940-68027940 on Build GRCh38
rs899789295 Chr.11: 68027592-68027592 on Build GRCh38
rs770131325 Chr.11: 68027882-68027882 on Build GRCh38
rs189012958 Chr.11: 68027593-68027593 on Build GRCh38
rs376190333 Chr.11: 68027878-68027878 on Build GRCh38
rs757480205 Chr.11: 68027983-68027983 on Build GRCh38
rs982907061 Chr.11: 68027809-68027809 on Build GRCh38
rs953576367 Chr.11: 68027812-68027812 on Build GRCh38
rs998621451 Chr.11: 68027637-68027637 on Build GRCh38
rs778923085 Chr.11: 68027700-68027700 on Build GRCh38
rs748162494 Chr.11: 68027872-68027872 on Build GRCh38
rs571804317 Chr.11: 68027847-68027847 on Build GRCh38
rs758506583 Chr.11: 68027873-68027873 on Build GRCh38
rs904366190 Chr.11: 68027862-68027862 on Build GRCh38
rs370859579 Chr.11: 68027881-68027881 on Build GRCh38
rs977402743 Chr.11: 68028026-68028026 on Build GRCh38
rs377025085 Chr.11: 68027820-68027820 on Build GRCh38
rs886701 Chr.11: 68027598-68027598 on Build GRCh38
rs763744773 Chr.11: 68027903-68027903 on Build GRCh38
rs369465283 Chr.11: 68027850-68027850 on Build GRCh38
rs753798617 Chr.11: 68027948-68027948 on Build GRCh38
rs965883993 Chr.11: 68027984-68027984 on Build GRCh38
rs777232783 Chr.11: 68027996-68027996 on Build GRCh38
rs762501177 Chr.11: 68027895-68027895 on Build GRCh38
rs1025208497 Chr.11: 68027708-68027708 on Build GRCh38
rs550673848 Chr.11: 68027866-68027866 on Build GRCh38
rs952921462 Chr.11: 68027898-68027898 on Build GRCh38
rs375874895 Chr.11: 68027806-68027806 on Build GRCh38
rs58238184 Chr.11: 68027913-68027914 on Build GRCh38
rs753889479 Chr.11: 68027907-68027908 on Build GRCh38
rs959748149 Chr.11: 68027672-68027672 on Build GRCh38
rs569239765 Chr.11: 68027867-68027867 on Build GRCh38
rs567436201 Chr.11: 68027977-68027977 on Build GRCh38
rs751776553 Chr.11: 68027982-68027982 on Build GRCh38
rs904271199 Chr.11: 68027611-68027611 on Build GRCh38
rs199635628 Chr.11: 68027838-68027838 on Build GRCh38
rs969594429 Chr.11: 68027880-68027880 on Build GRCh38
rs372813563 Chr.11: 68027819-68027819 on Build GRCh38
rs1021688203 Chr.11: 68028019-68028019 on Build GRCh38
rs980927437 Chr.11: 68027755-68027755 on Build GRCh38
rs866772584 Chr.11: 68027887-68027887 on Build GRCh38
rs749849588 Chr.11: 68028011-68028011 on Build GRCh38
rs3751082 Chr.11: 68027885-68027885 on Build GRCh38
rs766190163 Chr.11: 68027932-68027932 on Build GRCh38
rs866348707 Chr.11: 68027916-68027916 on Build GRCh38
rs374814356 Chr.11: 68027876-68027876 on Build GRCh38
rs532754681 Chr.11: 68027667-68027667 on Build GRCh38
rs372491942 Chr.11: 68027856-68027856 on Build GRCh38
rs777926357 Chr.11: 68027875-68027875 on Build GRCh38
rs563096242 Chr.11: 68028002-68028002 on Build GRCh38
rs754561141 Chr.11: 68027716-68027716 on Build GRCh38
rs760001254 Chr.11: 68027830-68027830 on Build GRCh38
rs377026164 Chr.11: 68027857-68027857 on Build GRCh38
rs1036303747 Chr.11: 68027905-68027905 on Build GRCh38
rs142085281 Chr.11: 68027989-68027989 on Build GRCh38
rs761437482 Chr.11: 68027904-68027904 on Build GRCh38
rs1041953185 Chr.11: 68027578-68027578 on Build GRCh38
rs950615309 Chr.11: 68027995-68027995 on Build GRCh38
rs144879452 Chr.11: 68027972-68027972 on Build GRCh38
rs549268172 Chr.11: 68027710-68027710 on Build GRCh38
rs779711812 Chr.11: 68027617-68027617 on Build GRCh38
rs369689965 Chr.11: 68027823-68027823 on Build GRCh38
rs369754167 Chr.11: 68027858-68027858 on Build GRCh38
rs767326025 Chr.11: 68027920-68027920 on Build GRCh38
rs757093900 Chr.11: 68027860-68027860 on Build GRCh38
rs534155304 Chr.11: 68027924-68027924 on Build GRCh38
rs887412878 Chr.11: 68027666-68027666 on Build GRCh38
rs772953072 Chr.11: 68028023-68028023 on Build GRCh38
rs968682341 Chr.11: 68027849-68027849 on Build GRCh38

View Full Product Details