Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68053951-68054452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKA
Entrez Gene ID
1119
Gene Name
choline kinase alpha
Gene Aliases
CHK, CK, CKI, EK
UniGene
Hs.77221
Gene Chromosome Location
Chr.11: 68052859-68121391 on Build GRCh38
Transcripts
NM_001277.2 NM_212469.1 XM_017017147.1 XM_017017148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676245 c.1351C>T p.Q451* Chr.11: 68054011-68054011 on Build GRCh38
931188 c.1334T>G p.F445C Chr.11: 68054028-68054028 on Build GRCh38
4036073 c.1321G>A p.A441T Chr.11: 68054041-68054041 on Build GRCh38
7335913 c.1324delC p.Q442fs*>16 Chr.11: 68054038-68054038 on Build GRCh38
4927951 c.1337A>G p.D446G Chr.11: 68054025-68054025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962796443 Chr.11: 68054212-68054212 on Build GRCh38
rs1012352194 Chr.11: 68054393-68054393 on Build GRCh38
rs573214593 Chr.11: 68054164-68054164 on Build GRCh38
rs994560889 Chr.11: 68054113-68054113 on Build GRCh38
rs987128827 Chr.11: 68054270-68054270 on Build GRCh38
rs549021507 Chr.11: 68054421-68054421 on Build GRCh38
rs759978491 Chr.11: 68054019-68054019 on Build GRCh38
rs962765311 Chr.11: 68054403-68054403 on Build GRCh38
rs771562960 Chr.11: 68054004-68054004 on Build GRCh38
rs141507470 Chr.11: 68054343-68054343 on Build GRCh38
rs771925607 Chr.11: 68054371-68054371 on Build GRCh38
rs747437768 Chr.11: 68053995-68053995 on Build GRCh38
rs773690486 Chr.11: 68053994-68053994 on Build GRCh38
rs1035156876 Chr.11: 68054158-68054158 on Build GRCh38
rs957378812 Chr.11: 68054197-68054197 on Build GRCh38
rs765206614 Chr.11: 68054002-68054004 on Build GRCh38
rs892781521 Chr.11: 68054102-68054102 on Build GRCh38
rs896629384 Chr.11: 68054392-68054392 on Build GRCh38
rs747241506 Chr.11: 68054099-68054099 on Build GRCh38
rs1033054010 Chr.11: 68054191-68054211 on Build GRCh38
rs772463621 Chr.11: 68053993-68053993 on Build GRCh38
rs778210484 Chr.11: 68054089-68054089 on Build GRCh38
rs757813958 Chr.11: 68054065-68054066 on Build GRCh38
rs35356476 Chr.11: 68054111-68054112 on Build GRCh38
rs918625118 Chr.11: 68054244-68054244 on Build GRCh38
rs186511614 Chr.11: 68054198-68054198 on Build GRCh38
rs951289912 Chr.11: 68054265-68054265 on Build GRCh38
rs560104032 Chr.11: 68054218-68054218 on Build GRCh38
rs377195521 Chr.11: 68054042-68054042 on Build GRCh38
rs1011526278 Chr.11: 68054366-68054366 on Build GRCh38
rs767302447 Chr.11: 68054058-68054058 on Build GRCh38
rs916216632 Chr.11: 68054252-68054252 on Build GRCh38
rs990766436 Chr.11: 68054246-68054247 on Build GRCh38
rs914314089 Chr.11: 68054302-68054302 on Build GRCh38
rs374879529 Chr.11: 68054118-68054118 on Build GRCh38
rs996249807 Chr.11: 68054191-68054206 on Build GRCh38
rs967419639 Chr.11: 68054376-68054376 on Build GRCh38
rs532886035 Chr.11: 68054275-68054275 on Build GRCh38
rs1020273308 Chr.11: 68054374-68054374 on Build GRCh38
rs942544183 Chr.11: 68054290-68054290 on Build GRCh38
rs748226236 Chr.11: 68054074-68054074 on Build GRCh38
rs1019144921 Chr.11: 68054149-68054149 on Build GRCh38
rs1023792722 Chr.11: 68054398-68054398 on Build GRCh38
rs1023694646 Chr.11: 68054207-68054207 on Build GRCh38
rs373511681 Chr.11: 68054084-68054084 on Build GRCh38
rs886181206 Chr.11: 68054288-68054288 on Build GRCh38
rs751338931 Chr.11: 68054052-68054052 on Build GRCh38
rs190925996 Chr.11: 68054418-68054418 on Build GRCh38
rs1041403187 Chr.11: 68054318-68054318 on Build GRCh38
rs966680489 Chr.11: 68054165-68054165 on Build GRCh38
rs1054305294 Chr.11: 68054316-68054316 on Build GRCh38
rs1048797214 Chr.11: 68054256-68054258 on Build GRCh38
rs961169935 Chr.11: 68054194-68054194 on Build GRCh38
rs539169773 Chr.11: 68054073-68054073 on Build GRCh38
rs999749712 Chr.11: 68054370-68054370 on Build GRCh38
rs561217210 Chr.11: 68054148-68054148 on Build GRCh38
rs140389865 Chr.11: 68054045-68054045 on Build GRCh38
rs575552565 Chr.11: 68054134-68054134 on Build GRCh38
rs191643946 Chr.11: 68054068-68054068 on Build GRCh38
rs764758799 Chr.11: 68054039-68054039 on Build GRCh38
rs551497216 Chr.11: 68054303-68054303 on Build GRCh38
rs763781221 Chr.11: 68054046-68054046 on Build GRCh38
rs777132759 Chr.11: 68054008-68054008 on Build GRCh38
rs1039579476 Chr.11: 68054292-68054292 on Build GRCh38
rs781746342 Chr.11: 68054073-68054074 on Build GRCh38
rs930297459 Chr.11: 68054253-68054253 on Build GRCh38
rs181456286 Chr.11: 68054145-68054145 on Build GRCh38
rs912368078 Chr.11: 68054283-68054283 on Build GRCh38
rs567571564 Chr.11: 68053977-68053977 on Build GRCh38
rs974036805 Chr.11: 68054216-68054216 on Build GRCh38
rs565506593 Chr.11: 68054251-68054251 on Build GRCh38
rs746555940 Chr.11: 68054320-68054320 on Build GRCh38
rs533414195 Chr.11: 68054394-68054394 on Build GRCh38
rs758224261 Chr.11: 68054120-68054124 on Build GRCh38
rs868722640 Chr.11: 68054308-68054308 on Build GRCh38
rs757108102 Chr.11: 68054054-68054054 on Build GRCh38
rs967471949 Chr.11: 68054211-68054211 on Build GRCh38

View Full Product Details