Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68061861-68062356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKA
Entrez Gene ID
1119
Gene Name
choline kinase alpha
Gene Aliases
CHK, CK, CKI, EK
UniGene
Hs.77221
Gene Chromosome Location
Chr.11: 68052859-68121391 on Build GRCh38
Transcripts
NM_001277.2 NM_212469.1 XM_017017147.1 XM_017017148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390045 c.1249C>A p.H417N Chr.11: 68062018-68062018 on Build GRCh38
3452397 c.1254C>T p.F418F Chr.11: 68062013-68062013 on Build GRCh38
931189 c.1287G>T p.K429N Chr.11: 68061980-68061980 on Build GRCh38
4808275 c.1299T>A p.I433I Chr.11: 68061968-68061968 on Build GRCh38
5477771 c.1290T>G p.I430M Chr.11: 68061977-68061977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199712734 Chr.11: 68062243-68062243 on Build GRCh38
rs200958186 Chr.11: 68061930-68061931 on Build GRCh38
rs767293195 Chr.11: 68061915-68061915 on Build GRCh38
rs772026034 Chr.11: 68062039-68062039 on Build GRCh38
rs971900472 Chr.11: 68062184-68062184 on Build GRCh38
rs200360093 Chr.11: 68061909-68061909 on Build GRCh38
rs567133469 Chr.11: 68062137-68062137 on Build GRCh38
rs761466150 Chr.11: 68062125-68062125 on Build GRCh38
rs765163323 Chr.11: 68062249-68062249 on Build GRCh38
rs897779982 Chr.11: 68061920-68061920 on Build GRCh38
rs751786298 Chr.11: 68061989-68061989 on Build GRCh38
rs557066871 Chr.11: 68062288-68062288 on Build GRCh38
rs761415538 Chr.11: 68062073-68062073 on Build GRCh38
rs778016291 Chr.11: 68061954-68061954 on Build GRCh38
rs569029223 Chr.11: 68062086-68062086 on Build GRCh38
rs1004477934 Chr.11: 68062196-68062196 on Build GRCh38
rs371329186 Chr.11: 68061937-68061937 on Build GRCh38
rs750881120 Chr.11: 68061911-68061911 on Build GRCh38
rs1015743364 Chr.11: 68062208-68062208 on Build GRCh38
rs768098030 Chr.11: 68062060-68062060 on Build GRCh38
rs548328747 Chr.11: 68062121-68062121 on Build GRCh38
rs757588762 Chr.11: 68061991-68061991 on Build GRCh38
rs538444800 Chr.11: 68062286-68062286 on Build GRCh38
rs376609261 Chr.11: 68061994-68061994 on Build GRCh38
rs749802170 Chr.11: 68062059-68062059 on Build GRCh38
rs1048760191 Chr.11: 68062179-68062179 on Build GRCh38
rs756537958 Chr.11: 68061919-68061920 on Build GRCh38
rs965421226 Chr.11: 68061912-68061912 on Build GRCh38
rs1022985374 Chr.11: 68062153-68062153 on Build GRCh38
rs767227890 Chr.11: 68062171-68062171 on Build GRCh38
rs904694282 Chr.11: 68062042-68062042 on Build GRCh38
rs888792981 Chr.11: 68062244-68062244 on Build GRCh38
rs770317844 Chr.11: 68062013-68062013 on Build GRCh38
rs17853642 Chr.11: 68062002-68062002 on Build GRCh38
rs143942851 Chr.11: 68062267-68062267 on Build GRCh38
rs1031922462 Chr.11: 68062076-68062076 on Build GRCh38
rs1015774590 Chr.11: 68061895-68061895 on Build GRCh38
rs1051399282 Chr.11: 68061928-68061928 on Build GRCh38
rs933255544 Chr.11: 68062200-68062200 on Build GRCh38
rs977256631 Chr.11: 68061921-68061921 on Build GRCh38
rs770973242 Chr.11: 68062178-68062178 on Build GRCh38
rs756056425 Chr.11: 68061925-68061925 on Build GRCh38
rs34788775 Chr.11: 68062178-68062179 on Build GRCh38
rs368949121 Chr.11: 68062113-68062113 on Build GRCh38
rs925868190 Chr.11: 68062010-68062010 on Build GRCh38
rs563358540 Chr.11: 68062195-68062195 on Build GRCh38
rs768889671 Chr.11: 68061920-68061924 on Build GRCh38
rs564974078 Chr.11: 68061922-68061922 on Build GRCh38
rs781585581 Chr.11: 68061992-68061992 on Build GRCh38
rs759396186 Chr.11: 68062230-68062230 on Build GRCh38
rs181422528 Chr.11: 68062289-68062289 on Build GRCh38
rs998390710 Chr.11: 68062281-68062281 on Build GRCh38
rs35303746 Chr.11: 68062197-68062198 on Build GRCh38
rs368441898 Chr.11: 68061931-68061931 on Build GRCh38
rs905641076 Chr.11: 68062176-68062176 on Build GRCh38
rs979130473 Chr.11: 68062335-68062335 on Build GRCh38
rs954169862 Chr.11: 68061979-68061979 on Build GRCh38
rs780401408 Chr.11: 68061920-68061920 on Build GRCh38
rs533201864 Chr.11: 68062182-68062185 on Build GRCh38
rs748302783 Chr.11: 68061931-68061931 on Build GRCh38
rs1051341839 Chr.11: 68062211-68062211 on Build GRCh38
rs773873878 Chr.11: 68062066-68062066 on Build GRCh38
rs773262565 Chr.11: 68062074-68062075 on Build GRCh38
rs368633943 Chr.11: 68062058-68062058 on Build GRCh38
rs747228424 Chr.11: 68062161-68062161 on Build GRCh38
rs1022179519 Chr.11: 68062041-68062041 on Build GRCh38
rs762776974 Chr.11: 68062316-68062316 on Build GRCh38

View Full Product Details