Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68685453-68685988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL
Entrez Gene ID
51083
Gene Name
galanin and GMAP prepropeptide
Gene Aliases
ETL8, GAL-GMAP, GALN, GLNN, GMAP
UniGene
Hs.278959
Gene Chromosome Location
Chr.11: 68684475-68691175 on Build GRCh38
Transcripts
NM_015973.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4940814 c.128T>C p.L43P Chr.11: 68685640-68685640 on Build GRCh38
690282 c.119G>T p.G40V Chr.11: 68685631-68685631 on Build GRCh38
4856407 c.115G>A p.A39T Chr.11: 68685627-68685627 on Build GRCh38
320541 c.102G>T p.W34C Chr.11: 68685614-68685614 on Build GRCh38
4884868 c.83C>A p.A28D Chr.11: 68685595-68685595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932410440 Chr.11: 68685903-68685903 on Build GRCh38
rs901354134 Chr.11: 68685764-68685764 on Build GRCh38
rs142908115 Chr.11: 68685773-68685773 on Build GRCh38
rs1021754734 Chr.11: 68685579-68685579 on Build GRCh38
rs1035857064 Chr.11: 68685822-68685822 on Build GRCh38
rs954064911 Chr.11: 68685502-68685502 on Build GRCh38
rs992688081 Chr.11: 68685859-68685859 on Build GRCh38
rs746516038 Chr.11: 68685626-68685626 on Build GRCh38
rs1015277127 Chr.11: 68685843-68685843 on Build GRCh38
rs756164130 Chr.11: 68685684-68685684 on Build GRCh38
rs750008754 Chr.11: 68685649-68685650 on Build GRCh38
rs939664923 Chr.11: 68685962-68685962 on Build GRCh38
rs1008673104 Chr.11: 68685525-68685525 on Build GRCh38
rs915728995 Chr.11: 68685934-68685934 on Build GRCh38
rs575522673 Chr.11: 68685854-68685854 on Build GRCh38
rs1036380082 Chr.11: 68685632-68685632 on Build GRCh38
rs770751421 Chr.11: 68685627-68685627 on Build GRCh38
rs970169359 Chr.11: 68685606-68685606 on Build GRCh38
rs571783123 Chr.11: 68685834-68685834 on Build GRCh38
rs915234224 Chr.11: 68685519-68685519 on Build GRCh38
rs138422737 Chr.11: 68685744-68685744 on Build GRCh38
rs550985433 Chr.11: 68685726-68685726 on Build GRCh38
rs973837914 Chr.11: 68685851-68685851 on Build GRCh38
rs919771201 Chr.11: 68685723-68685723 on Build GRCh38
rs865991869 Chr.11: 68685696-68685696 on Build GRCh38
rs567271827 Chr.11: 68685779-68685779 on Build GRCh38
rs759431773 Chr.11: 68685636-68685636 on Build GRCh38
rs948514091 Chr.11: 68685947-68685947 on Build GRCh38
rs540382719 Chr.11: 68685689-68685689 on Build GRCh38
rs764086315 Chr.11: 68685658-68685658 on Build GRCh38
rs891070816 Chr.11: 68685743-68685743 on Build GRCh38
rs888117760 Chr.11: 68685732-68685732 on Build GRCh38
rs919702597 Chr.11: 68685872-68685872 on Build GRCh38
rs575560342 Chr.11: 68685551-68685551 on Build GRCh38
rs1057517661 Chr.11: 68685628-68685628 on Build GRCh38
rs12289299 Chr.11: 68685850-68685850 on Build GRCh38
rs960609260 Chr.11: 68685833-68685833 on Build GRCh38
rs772807519 Chr.11: 68685629-68685629 on Build GRCh38
rs1049568745 Chr.11: 68685727-68685727 on Build GRCh38
rs1011575430 Chr.11: 68685573-68685573 on Build GRCh38
rs749701826 Chr.11: 68685600-68685600 on Build GRCh38
rs985736095 Chr.11: 68685932-68685932 on Build GRCh38
rs557564546 Chr.11: 68685853-68685853 on Build GRCh38
rs762854158 Chr.11: 68685651-68685651 on Build GRCh38
rs542594677 Chr.11: 68685595-68685595 on Build GRCh38
rs775508840 Chr.11: 68685649-68685649 on Build GRCh38
rs377221826 Chr.11: 68685692-68685692 on Build GRCh38
rs772658511 Chr.11: 68685623-68685623 on Build GRCh38
rs750508297 Chr.11: 68685662-68685662 on Build GRCh38
rs912358626 Chr.11: 68685951-68685951 on Build GRCh38
rs939755748 Chr.11: 68685699-68685699 on Build GRCh38
rs998287274 Chr.11: 68685772-68685772 on Build GRCh38
rs960308302 Chr.11: 68685666-68685666 on Build GRCh38
rs1048559353 Chr.11: 68685731-68685731 on Build GRCh38
rs757360072 Chr.11: 68685549-68685549 on Build GRCh38
rs370408784 Chr.11: 68685567-68685567 on Build GRCh38
rs143418460 Chr.11: 68685788-68685788 on Build GRCh38
rs3136537 Chr.11: 68685961-68685961 on Build GRCh38
rs921309431 Chr.11: 68685478-68685478 on Build GRCh38
rs183547277 Chr.11: 68685700-68685700 on Build GRCh38
rs891762641 Chr.11: 68685774-68685774 on Build GRCh38
rs528520052 Chr.11: 68685618-68685618 on Build GRCh38
rs977742548 Chr.11: 68685837-68685837 on Build GRCh38
rs779506188 Chr.11: 68685601-68685601 on Build GRCh38
rs201001800 Chr.11: 68685693-68685693 on Build GRCh38
rs542917636 Chr.11: 68685952-68685952 on Build GRCh38
rs694066 Chr.11: 68685517-68685517 on Build GRCh38
rs578114657 Chr.11: 68685487-68685487 on Build GRCh38
rs1011627651 Chr.11: 68685778-68685778 on Build GRCh38
rs199667492 Chr.11: 68685546-68685546 on Build GRCh38
rs148372434 Chr.11: 68685807-68685807 on Build GRCh38
rs879201115 Chr.11: 68685491-68685491 on Build GRCh38
rs756393174 Chr.11: 68685566-68685566 on Build GRCh38
rs530344730 Chr.11: 68685643-68685643 on Build GRCh38
rs551583863 Chr.11: 68685709-68685709 on Build GRCh38
rs201520007 Chr.11: 68685616-68685616 on Build GRCh38
rs750501090 Chr.11: 68685561-68685561 on Build GRCh38
rs766611326 Chr.11: 68685688-68685688 on Build GRCh38
rs905940489 Chr.11: 68685806-68685806 on Build GRCh38

View Full Product Details