Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69773321-69773813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF4
Entrez Gene ID
2249
Gene Name
fibroblast growth factor 4
Gene Aliases
HBGF-4, HST, HST-1, HSTF1, K-FGF, KFGF
UniGene
Hs.1755
Gene Chromosome Location
Chr.11: 69773029-69775403 on Build GRCh38
Transcripts
NM_002007.2 XM_005273847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3967802 c.506A>C p.Y169S Chr.11: 69773424-69773424 on Build GRCh38
6462021 c.564G>A p.K188K Chr.11: 69773366-69773366 on Build GRCh38
3980632 c.504C>T p.A168A Chr.11: 69773426-69773426 on Build GRCh38
6537431 c.462G>C p.E154D Chr.11: 69773468-69773468 on Build GRCh38
6981230 c.591G>A p.M197I Chr.11: 69773339-69773339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763643003 Chr.11: 69773447-69773447 on Build GRCh38
rs386754668 Chr.11: 69773449-69773450 on Build GRCh38
rs920894821 Chr.11: 69773770-69773770 on Build GRCh38
rs894254552 Chr.11: 69773664-69773664 on Build GRCh38
rs930844659 Chr.11: 69773430-69773430 on Build GRCh38
rs769378385 Chr.11: 69773423-69773423 on Build GRCh38
rs535935656 Chr.11: 69773429-69773429 on Build GRCh38
rs756008893 Chr.11: 69773466-69773466 on Build GRCh38
rs138379196 Chr.11: 69773521-69773521 on Build GRCh38
rs140780726 Chr.11: 69773393-69773393 on Build GRCh38
rs537673069 Chr.11: 69773638-69773638 on Build GRCh38
rs775122084 Chr.11: 69773533-69773533 on Build GRCh38
rs1026778128 Chr.11: 69773567-69773567 on Build GRCh38
rs992524703 Chr.11: 69773398-69773398 on Build GRCh38
rs752416500 Chr.11: 69773347-69773347 on Build GRCh38
rs145463849 Chr.11: 69773450-69773450 on Build GRCh38
rs908478812 Chr.11: 69773665-69773665 on Build GRCh38
rs748019249 Chr.11: 69773359-69773360 on Build GRCh38
rs971752012 Chr.11: 69773739-69773739 on Build GRCh38
rs533842802 Chr.11: 69773473-69773473 on Build GRCh38
rs982623064 Chr.11: 69773658-69773658 on Build GRCh38
rs780987779 Chr.11: 69773353-69773359 on Build GRCh38
rs748749676 Chr.11: 69773623-69773623 on Build GRCh38
rs143738659 Chr.11: 69773405-69773405 on Build GRCh38
rs753962702 Chr.11: 69773381-69773381 on Build GRCh38
rs755033485 Chr.11: 69773390-69773390 on Build GRCh38
rs758826241 Chr.11: 69773348-69773348 on Build GRCh38
rs781693830 Chr.11: 69773534-69773534 on Build GRCh38
rs986306143 Chr.11: 69773434-69773434 on Build GRCh38
rs757859717 Chr.11: 69773499-69773499 on Build GRCh38
rs1024034666 Chr.11: 69773690-69773690 on Build GRCh38
rs866953016 Chr.11: 69773361-69773361 on Build GRCh38
rs976859379 Chr.11: 69773697-69773697 on Build GRCh38
rs200135119 Chr.11: 69773462-69773462 on Build GRCh38
rs961706373 Chr.11: 69773721-69773721 on Build GRCh38
rs751257747 Chr.11: 69773355-69773355 on Build GRCh38
rs200919082 Chr.11: 69773488-69773488 on Build GRCh38
rs759166949 Chr.11: 69773408-69773408 on Build GRCh38
rs779058257 Chr.11: 69773469-69773469 on Build GRCh38
rs780362260 Chr.11: 69773359-69773359 on Build GRCh38
rs781106458 Chr.11: 69773364-69773364 on Build GRCh38
rs758706825 Chr.11: 69773474-69773474 on Build GRCh38
rs763955342 Chr.11: 69773349-69773349 on Build GRCh38
rs556304786 Chr.11: 69773728-69773728 on Build GRCh38
rs140567674 Chr.11: 69773449-69773449 on Build GRCh38
rs1028355170 Chr.11: 69773762-69773762 on Build GRCh38
rs772115968 Chr.11: 69773403-69773403 on Build GRCh38
rs150074947 Chr.11: 69773407-69773407 on Build GRCh38
rs746380967 Chr.11: 69773515-69773515 on Build GRCh38
rs550329316 Chr.11: 69773404-69773404 on Build GRCh38
rs1000471491 Chr.11: 69773582-69773582 on Build GRCh38
rs112765358 Chr.11: 69773453-69773453 on Build GRCh38
rs576648614 Chr.11: 69773545-69773545 on Build GRCh38
rs781654973 Chr.11: 69773504-69773504 on Build GRCh38
rs748429697 Chr.11: 69773394-69773394 on Build GRCh38
rs201349393 Chr.11: 69773428-69773428 on Build GRCh38
rs1034175751 Chr.11: 69773651-69773651 on Build GRCh38
rs574486116 Chr.11: 69773779-69773779 on Build GRCh38
rs1042028355 Chr.11: 69773479-69773479 on Build GRCh38
rs1014033515 Chr.11: 69773673-69773673 on Build GRCh38
rs762363344 Chr.11: 69773536-69773536 on Build GRCh38
rs750386794 Chr.11: 69773463-69773463 on Build GRCh38

View Full Product Details