Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73310327-73310821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF17
Entrez Gene ID
9828
Gene Name
Rho guanine nucleotide exchange factor 17
Gene Aliases
P164RHOGEF, TEM4, p164-RhoGEF
UniGene
Hs.533719
Gene Chromosome Location
Chr.11: 73305961-73369380 on Build GRCh38
Transcripts
NM_014786.3 XM_017018623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2016851 c.1844delC p.P617fs*10 Chr.11: 73310482-73310482 on Build GRCh38
6604040 c.1759G>A p.V587I Chr.11: 73310397-73310397 on Build GRCh38
3359389 c.1984C>A p.L662M Chr.11: 73310622-73310622 on Build GRCh38
2016863 c.2161G>A p.E721K Chr.11: 73310799-73310799 on Build GRCh38
3810384 c.2154T>G p.G718G Chr.11: 73310792-73310792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747752927 Chr.11: 73310397-73310397 on Build GRCh38
rs367623424 Chr.11: 73310684-73310684 on Build GRCh38
rs143223501 Chr.11: 73310666-73310666 on Build GRCh38
rs762014607 Chr.11: 73310372-73310372 on Build GRCh38
rs756814001 Chr.11: 73310581-73310581 on Build GRCh38
rs370293085 Chr.11: 73310365-73310365 on Build GRCh38
rs987216909 Chr.11: 73310769-73310769 on Build GRCh38
rs763404198 Chr.11: 73310659-73310659 on Build GRCh38
rs374814851 Chr.11: 73310671-73310671 on Build GRCh38
rs753032222 Chr.11: 73310473-73310473 on Build GRCh38
rs1055250437 Chr.11: 73310656-73310656 on Build GRCh38
rs753672523 Chr.11: 73310695-73310695 on Build GRCh38
rs762386120 Chr.11: 73310760-73310761 on Build GRCh38
rs755362750 Chr.11: 73310466-73310466 on Build GRCh38
rs763803068 Chr.11: 73310571-73310571 on Build GRCh38
rs745949382 Chr.11: 73310721-73310721 on Build GRCh38
rs148112775 Chr.11: 73310709-73310709 on Build GRCh38
rs780614742 Chr.11: 73310601-73310601 on Build GRCh38
rs574600996 Chr.11: 73310354-73310354 on Build GRCh38
rs765725022 Chr.11: 73310556-73310556 on Build GRCh38
rs138410023 Chr.11: 73310417-73310417 on Build GRCh38
rs772693765 Chr.11: 73310758-73310758 on Build GRCh38
rs149213151 Chr.11: 73310449-73310449 on Build GRCh38
rs756180677 Chr.11: 73310618-73310618 on Build GRCh38
rs747975543 Chr.11: 73310706-73310706 on Build GRCh38
rs749967997 Chr.11: 73310678-73310678 on Build GRCh38
rs749083960 Chr.11: 73310519-73310519 on Build GRCh38
rs766325143 Chr.11: 73310544-73310544 on Build GRCh38
rs376001603 Chr.11: 73310680-73310680 on Build GRCh38
rs749871258 Chr.11: 73310759-73310760 on Build GRCh38
rs1048543442 Chr.11: 73310707-73310707 on Build GRCh38
rs994320620 Chr.11: 73310698-73310698 on Build GRCh38
rs199547585 Chr.11: 73310486-73310486 on Build GRCh38
rs373850527 Chr.11: 73310775-73310775 on Build GRCh38
rs200616023 Chr.11: 73310421-73310421 on Build GRCh38
rs780084735 Chr.11: 73310619-73310619 on Build GRCh38
rs369681028 Chr.11: 73310757-73310757 on Build GRCh38
rs774644446 Chr.11: 73310736-73310736 on Build GRCh38
rs769174361 Chr.11: 73310734-73310734 on Build GRCh38
rs751131302 Chr.11: 73310580-73310580 on Build GRCh38
rs925062213 Chr.11: 73310640-73310640 on Build GRCh38
rs567330760 Chr.11: 73310487-73310487 on Build GRCh38
rs1054798573 Chr.11: 73310358-73310358 on Build GRCh38
rs769100884 Chr.11: 73310799-73310799 on Build GRCh38
rs773242491 Chr.11: 73310524-73310524 on Build GRCh38
rs376978053 Chr.11: 73310348-73310348 on Build GRCh38
rs764421666 Chr.11: 73310363-73310363 on Build GRCh38
rs781220764 Chr.11: 73310485-73310485 on Build GRCh38
rs760079262 Chr.11: 73310553-73310553 on Build GRCh38
rs903590448 Chr.11: 73310352-73310352 on Build GRCh38
rs759387116 Chr.11: 73310359-73310359 on Build GRCh38
rs200068545 Chr.11: 73310742-73310742 on Build GRCh38
rs139620086 Chr.11: 73310753-73310753 on Build GRCh38
rs745662326 Chr.11: 73310424-73310424 on Build GRCh38
rs749381710 Chr.11: 73310798-73310798 on Build GRCh38
rs949052094 Chr.11: 73310668-73310668 on Build GRCh38
rs772961945 Chr.11: 73310744-73310744 on Build GRCh38
rs776672879 Chr.11: 73310549-73310549 on Build GRCh38
rs760843345 Chr.11: 73310531-73310531 on Build GRCh38
rs754262806 Chr.11: 73310462-73310462 on Build GRCh38
rs766038618 Chr.11: 73310752-73310752 on Build GRCh38
rs747249002 Chr.11: 73310643-73310643 on Build GRCh38
rs1042684194 Chr.11: 73310583-73310583 on Build GRCh38
rs777693036 Chr.11: 73310444-73310444 on Build GRCh38
rs777379807 Chr.11: 73310400-73310400 on Build GRCh38
rs756713299 Chr.11: 73310381-73310381 on Build GRCh38
rs150609147 Chr.11: 73310754-73310754 on Build GRCh38
rs760614357 Chr.11: 73310457-73310457 on Build GRCh38
rs775464904 Chr.11: 73310496-73310496 on Build GRCh38
rs371501281 Chr.11: 73310697-73310697 on Build GRCh38
rs963268019 Chr.11: 73310408-73310408 on Build GRCh38
rs777275125 Chr.11: 73310481-73310481 on Build GRCh38
rs558565073 Chr.11: 73310726-73310726 on Build GRCh38
rs771112527 Chr.11: 73310708-73310708 on Build GRCh38
rs369875415 Chr.11: 73310625-73310625 on Build GRCh38
rs774734722 Chr.11: 73310425-73310425 on Build GRCh38
rs760284970 Chr.11: 73310751-73310751 on Build GRCh38
rs756651626 Chr.11: 73310483-73310483 on Build GRCh38
rs146910724 Chr.11: 73310468-73310468 on Build GRCh38
rs775904992 Chr.11: 73310654-73310654 on Build GRCh38
rs749921006 Chr.11: 73310611-73310611 on Build GRCh38
rs765641252 Chr.11: 73310467-73310467 on Build GRCh38
rs370888783 Chr.11: 73310459-73310459 on Build GRCh38
rs771155882 Chr.11: 73310644-73310644 on Build GRCh38
rs373017048 Chr.11: 73310728-73310728 on Build GRCh38
rs554558080 Chr.11: 73310788-73310788 on Build GRCh38
rs140800120 Chr.11: 73310357-73310357 on Build GRCh38
rs763165327 Chr.11: 73310569-73310569 on Build GRCh38
rs778572734 Chr.11: 73310391-73310391 on Build GRCh38
rs754243792 Chr.11: 73310384-73310384 on Build GRCh38
rs780146576 Chr.11: 73310797-73310797 on Build GRCh38
rs1027057021 Chr.11: 73310456-73310456 on Build GRCh38
rs745438319 Chr.11: 73310514-73310514 on Build GRCh38
rs761146124 Chr.11: 73310481-73310482 on Build GRCh38
rs759596749 Chr.11: 73310646-73310646 on Build GRCh38
rs770955437 Chr.11: 73310419-73310419 on Build GRCh38
rs779975857 Chr.11: 73310647-73310647 on Build GRCh38
rs762173891 Chr.11: 73310430-73310430 on Build GRCh38
rs781420908 Chr.11: 73310779-73310779 on Build GRCh38
rs540392597 Chr.11: 73310355-73310355 on Build GRCh38
rs139212193 Chr.11: 73310523-73310523 on Build GRCh38
rs776761143 Chr.11: 73310645-73310645 on Build GRCh38
rs200638279 Chr.11: 73310452-73310452 on Build GRCh38
rs764314700 Chr.11: 73310662-73310662 on Build GRCh38
rs776553502 Chr.11: 73310356-73310356 on Build GRCh38
rs756305140 Chr.11: 73310791-73310791 on Build GRCh38
rs143536153 Chr.11: 73310766-73310766 on Build GRCh38
rs746538707 Chr.11: 73310482-73310482 on Build GRCh38
rs867371892 Chr.11: 73310534-73310534 on Build GRCh38
rs191562625 Chr.11: 73310558-73310558 on Build GRCh38
rs147471306 Chr.11: 73310672-73310672 on Build GRCh38
rs764619467 Chr.11: 73310489-73310489 on Build GRCh38
rs1018751527 Chr.11: 73310725-73310725 on Build GRCh38
rs758109677 Chr.11: 73310477-73310477 on Build GRCh38
rs778618235 Chr.11: 73310699-73310699 on Build GRCh38
rs751928355 Chr.11: 73310664-73310664 on Build GRCh38
rs755272795 Chr.11: 73310390-73310390 on Build GRCh38
rs766783518 Chr.11: 73310383-73310383 on Build GRCh38
rs267603181 Chr.11: 73310774-73310774 on Build GRCh38
rs149321700 Chr.11: 73310373-73310373 on Build GRCh38
rs749100386 Chr.11: 73310620-73310620 on Build GRCh38
rs778083149 Chr.11: 73310631-73310631 on Build GRCh38

View Full Product Details