Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73409463-73409965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168A
Entrez Gene ID
23201
Gene Name
family with sequence similarity 168 member A
Gene Aliases
KIAA0280, TCRP1
UniGene
Hs.475334
Gene Chromosome Location
Chr.11: 73400478-73598189 on Build GRCh38
Transcripts
NM_001286050.1 NM_001286051.1 NM_015159.2 XM_017017415.1 XM_017017416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206179 c.421-1G>T p.? Chr.11: 73409662-73409662 on Build GRCh38
4036603 c.531C>T p.A177A Chr.11: 73409551-73409551 on Build GRCh38
1251804 c.528T>G p.V176V Chr.11: 73409554-73409554 on Build GRCh38
6658846 c.498C>T p.S166S Chr.11: 73409584-73409584 on Build GRCh38
6249297 c.490C>A p.P164T Chr.11: 73409592-73409592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961342568 Chr.11: 73409749-73409749 on Build GRCh38
rs765156502 Chr.11: 73409556-73409556 on Build GRCh38
rs369637071 Chr.11: 73409536-73409536 on Build GRCh38
rs189657156 Chr.11: 73409763-73409763 on Build GRCh38
rs762916579 Chr.11: 73409696-73409696 on Build GRCh38
rs777496675 Chr.11: 73409603-73409603 on Build GRCh38
rs904295579 Chr.11: 73409620-73409620 on Build GRCh38
rs945687137 Chr.11: 73409842-73409842 on Build GRCh38
rs1050553228 Chr.11: 73409721-73409721 on Build GRCh38
rs10793060 Chr.11: 73409608-73409608 on Build GRCh38
rs985601529 Chr.11: 73409796-73409796 on Build GRCh38
rs911371469 Chr.11: 73409812-73409812 on Build GRCh38
rs372550617 Chr.11: 73409550-73409550 on Build GRCh38
rs1014859983 Chr.11: 73409728-73409728 on Build GRCh38
rs575303371 Chr.11: 73409552-73409552 on Build GRCh38
rs764958504 Chr.11: 73409884-73409884 on Build GRCh38
rs763995542 Chr.11: 73409697-73409697 on Build GRCh38
rs201339524 Chr.11: 73409675-73409675 on Build GRCh38
rs767930955 Chr.11: 73409545-73409545 on Build GRCh38
rs560944463 Chr.11: 73409714-73409714 on Build GRCh38
rs373487023 Chr.11: 73409707-73409707 on Build GRCh38
rs560062943 Chr.11: 73409781-73409781 on Build GRCh38
rs761385621 Chr.11: 73409792-73409792 on Build GRCh38
rs749363209 Chr.11: 73409529-73409529 on Build GRCh38
rs957771518 Chr.11: 73409863-73409863 on Build GRCh38
rs746033767 Chr.11: 73409503-73409503 on Build GRCh38
rs754029318 Chr.11: 73409706-73409706 on Build GRCh38
rs573306231 Chr.11: 73409830-73409830 on Build GRCh38
rs907832379 Chr.11: 73409509-73409509 on Build GRCh38
rs944283771 Chr.11: 73409543-73409543 on Build GRCh38
rs780308609 Chr.11: 73409617-73409617 on Build GRCh38
rs182735953 Chr.11: 73409829-73409829 on Build GRCh38
rs749394184 Chr.11: 73409621-73409621 on Build GRCh38
rs761937954 Chr.11: 73409551-73409551 on Build GRCh38
rs1032031144 Chr.11: 73409843-73409843 on Build GRCh38
rs758974385 Chr.11: 73409687-73409687 on Build GRCh38
rs772593525 Chr.11: 73409647-73409647 on Build GRCh38
rs1014581706 Chr.11: 73409708-73409708 on Build GRCh38
rs768712052 Chr.11: 73409623-73409623 on Build GRCh38
rs775472630 Chr.11: 73409527-73409527 on Build GRCh38
rs781398055 Chr.11: 73409497-73409497 on Build GRCh38
rs891427523 Chr.11: 73409580-73409580 on Build GRCh38
rs760651505 Chr.11: 73409638-73409638 on Build GRCh38
rs764748428 Chr.11: 73409688-73409688 on Build GRCh38
rs553526282 Chr.11: 73409912-73409912 on Build GRCh38
rs373720143 Chr.11: 73409641-73409641 on Build GRCh38
rs774732056 Chr.11: 73409636-73409636 on Build GRCh38
rs1034580294 Chr.11: 73409692-73409692 on Build GRCh38
rs769099169 Chr.11: 73409534-73409534 on Build GRCh38
rs769891807 Chr.11: 73409525-73409525 on Build GRCh38
rs937085767 Chr.11: 73409929-73409929 on Build GRCh38
rs944227369 Chr.11: 73409813-73409813 on Build GRCh38
rs773017401 Chr.11: 73409546-73409546 on Build GRCh38
rs757070011 Chr.11: 73409701-73409701 on Build GRCh38
rs760366731 Chr.11: 73409548-73409548 on Build GRCh38
rs368988014 Chr.11: 73409694-73409694 on Build GRCh38
rs574619826 Chr.11: 73409750-73409750 on Build GRCh38
rs746774463 Chr.11: 73409605-73409605 on Build GRCh38
rs750104199 Chr.11: 73409890-73409890 on Build GRCh38
rs758203371 Chr.11: 73409602-73409602 on Build GRCh38
rs952887187 Chr.11: 73409786-73409786 on Build GRCh38
rs758187045 Chr.11: 73409487-73409487 on Build GRCh38
rs377034384 Chr.11: 73409599-73409599 on Build GRCh38
rs773755829 Chr.11: 73409654-73409654 on Build GRCh38
rs774913792 Chr.11: 73409535-73409535 on Build GRCh38

View Full Product Details