Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73411239-73411733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168A
Entrez Gene ID
23201
Gene Name
family with sequence similarity 168 member A
Gene Aliases
KIAA0280, TCRP1
UniGene
Hs.475334
Gene Chromosome Location
Chr.11: 73400478-73598189 on Build GRCh38
Transcripts
NM_001286050.1 NM_001286051.1 NM_015159.2 XM_017017415.1 XM_017017416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658848 c.368C>T p.T123M Chr.11: 73411446-73411446 on Build GRCh38
1356885 c.339delC p.Y114fs*89 Chr.11: 73411475-73411475 on Build GRCh38
5983961 c.402G>A p.Q134Q Chr.11: 73411412-73411412 on Build GRCh38
383066 c.311C>T p.P104L Chr.11: 73411503-73411503 on Build GRCh38
4832043 c.384C>G p.I128M Chr.11: 73411430-73411430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998866608 Chr.11: 73411496-73411496 on Build GRCh38
rs888970559 Chr.11: 73411380-73411380 on Build GRCh38
rs759549715 Chr.11: 73411263-73411265 on Build GRCh38
rs769219706 Chr.11: 73411425-73411425 on Build GRCh38
rs768093800 Chr.11: 73411508-73411508 on Build GRCh38
rs993778735 Chr.11: 73411621-73411621 on Build GRCh38
rs555909581 Chr.11: 73411279-73411279 on Build GRCh38
rs764085458 Chr.11: 73411447-73411447 on Build GRCh38
rs778982542 Chr.11: 73411617-73411617 on Build GRCh38
rs767549002 Chr.11: 73411353-73411355 on Build GRCh38
rs774290233 Chr.11: 73411515-73411515 on Build GRCh38
rs550319956 Chr.11: 73411704-73411704 on Build GRCh38
rs188939288 Chr.11: 73411329-73411329 on Build GRCh38
rs756514277 Chr.11: 73411321-73411321 on Build GRCh38
rs1938777 Chr.11: 73411295-73411295 on Build GRCh38
rs772757315 Chr.11: 73411363-73411363 on Build GRCh38
rs918931903 Chr.11: 73411711-73411711 on Build GRCh38
rs773112252 Chr.11: 73411553-73411553 on Build GRCh38
rs771261861 Chr.11: 73411390-73411390 on Build GRCh38
rs548487821 Chr.11: 73411322-73411322 on Build GRCh38
rs755265980 Chr.11: 73411475-73411475 on Build GRCh38
rs778429517 Chr.11: 73411487-73411487 on Build GRCh38
rs910381846 Chr.11: 73411316-73411316 on Build GRCh38
rs951839734 Chr.11: 73411654-73411654 on Build GRCh38
rs1054952565 Chr.11: 73411443-73411443 on Build GRCh38
rs146119169 Chr.11: 73411262-73411262 on Build GRCh38
rs781581760 Chr.11: 73411495-73411495 on Build GRCh38
rs375367550 Chr.11: 73411446-73411446 on Build GRCh38
rs752903882 Chr.11: 73411575-73411575 on Build GRCh38
rs527683756 Chr.11: 73411432-73411432 on Build GRCh38
rs761523348 Chr.11: 73411455-73411455 on Build GRCh38
rs748210527 Chr.11: 73411356-73411356 on Build GRCh38
rs754039172 Chr.11: 73411376-73411376 on Build GRCh38
rs200934649 Chr.11: 73411503-73411503 on Build GRCh38
rs747448032 Chr.11: 73411374-73411374 on Build GRCh38
rs561690152 Chr.11: 73411342-73411342 on Build GRCh38
rs1005928621 Chr.11: 73411690-73411690 on Build GRCh38
rs940288687 Chr.11: 73411694-73411694 on Build GRCh38
rs765627065 Chr.11: 73411561-73411561 on Build GRCh38
rs767324110 Chr.11: 73411550-73411550 on Build GRCh38
rs767289245 Chr.11: 73411460-73411460 on Build GRCh38
rs1026009157 Chr.11: 73411273-73411273 on Build GRCh38
rs533431460 Chr.11: 73411585-73411585 on Build GRCh38
rs1023047934 Chr.11: 73411624-73411624 on Build GRCh38
rs747462016 Chr.11: 73411490-73411490 on Build GRCh38
rs376542756 Chr.11: 73411556-73411556 on Build GRCh38
rs531063471 Chr.11: 73411532-73411532 on Build GRCh38
rs768153622 Chr.11: 73411274-73411274 on Build GRCh38
rs377443780 Chr.11: 73411351-73411351 on Build GRCh38
rs752885398 Chr.11: 73411478-73411478 on Build GRCh38
rs751430743 Chr.11: 73411448-73411448 on Build GRCh38
rs903228678 Chr.11: 73411373-73411373 on Build GRCh38
rs748800138 Chr.11: 73411507-73411507 on Build GRCh38
rs928900784 Chr.11: 73411680-73411680 on Build GRCh38
rs778415171 Chr.11: 73411372-73411372 on Build GRCh38
rs769542145 Chr.11: 73411502-73411502 on Build GRCh38
rs752955985 Chr.11: 73411373-73411380 on Build GRCh38
rs1013515906 Chr.11: 73411545-73411545 on Build GRCh38
rs758551943 Chr.11: 73411483-73411483 on Build GRCh38
rs981609469 Chr.11: 73411653-73411654 on Build GRCh38
rs376201217 Chr.11: 73411476-73411476 on Build GRCh38
rs756326960 Chr.11: 73411528-73411528 on Build GRCh38
rs771224666 Chr.11: 73411494-73411494 on Build GRCh38
rs939023074 Chr.11: 73411324-73411324 on Build GRCh38
rs564411184 Chr.11: 73411526-73411526 on Build GRCh38
rs776161582 Chr.11: 73411403-73411403 on Build GRCh38
rs754035792 Chr.11: 73411472-73411472 on Build GRCh38
rs970206750 Chr.11: 73411625-73411625 on Build GRCh38
rs200603016 Chr.11: 73411412-73411412 on Build GRCh38
rs1025976978 Chr.11: 73411631-73411631 on Build GRCh38

View Full Product Details