Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73958238-73958745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB13
Entrez Gene ID
374407
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Gene Aliases
RSPH16A, TSARG5, TSARG6
UniGene
Hs.567888
Gene Chromosome Location
Chr.11: 73950319-73970287 on Build GRCh38
Transcripts
NM_153614.3 XM_005273984.3 XM_011545004.2 XM_011545005.2 XM_011545007.2 XM_011545009.2 XM_011545013.2 XM_011545014.2 XM_017017675.1 XM_017017676.1 XM_017017677.1 XM_017017678.1 XM_017017679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7115474 c.77G>A p.R26K Chr.11: 73958325-73958325 on Build GRCh38
2017154 c.70T>C p.Y24H Chr.11: 73958318-73958318 on Build GRCh38
4574788 c.156C>T p.Y52Y Chr.11: 73958404-73958404 on Build GRCh38
6358785 c.172+1G>C p.? Chr.11: 73958421-73958421 on Build GRCh38
4678661 c.90G>T p.K30N Chr.11: 73958338-73958338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201516616 Chr.11: 73958409-73958409 on Build GRCh38
rs763216106 Chr.11: 73958292-73958292 on Build GRCh38
rs761885163 Chr.11: 73958391-73958391 on Build GRCh38
rs916041188 Chr.11: 73958675-73958675 on Build GRCh38
rs757937417 Chr.11: 73958347-73958347 on Build GRCh38
rs752395903 Chr.11: 73958448-73958448 on Build GRCh38
rs776401769 Chr.11: 73958290-73958290 on Build GRCh38
rs112339068 Chr.11: 73958706-73958706 on Build GRCh38
rs183789783 Chr.11: 73958572-73958572 on Build GRCh38
rs891151205 Chr.11: 73958639-73958639 on Build GRCh38
rs375603870 Chr.11: 73958321-73958321 on Build GRCh38
rs368039225 Chr.11: 73958428-73958428 on Build GRCh38
rs146073659 Chr.11: 73958404-73958404 on Build GRCh38
rs149369852 Chr.11: 73958329-73958329 on Build GRCh38
rs370839231 Chr.11: 73958280-73958280 on Build GRCh38
rs374177973 Chr.11: 73958283-73958283 on Build GRCh38
rs894652650 Chr.11: 73958315-73958315 on Build GRCh38
rs750954470 Chr.11: 73958317-73958317 on Build GRCh38
rs538182581 Chr.11: 73958499-73958499 on Build GRCh38
rs1047391227 Chr.11: 73958718-73958718 on Build GRCh38
rs754716956 Chr.11: 73958327-73958327 on Build GRCh38
rs771441391 Chr.11: 73958281-73958281 on Build GRCh38
rs767449214 Chr.11: 73958314-73958314 on Build GRCh38
rs748673996 Chr.11: 73958347-73958347 on Build GRCh38
rs567188885 Chr.11: 73958506-73958506 on Build GRCh38
rs548844823 Chr.11: 73958500-73958500 on Build GRCh38
rs1029773024 Chr.11: 73958544-73958544 on Build GRCh38
rs941208498 Chr.11: 73958574-73958574 on Build GRCh38
rs746415739 Chr.11: 73958259-73958259 on Build GRCh38
rs990430808 Chr.11: 73958671-73958671 on Build GRCh38
rs774459652 Chr.11: 73958306-73958306 on Build GRCh38
rs768857246 Chr.11: 73958376-73958376 on Build GRCh38
rs747404191 Chr.11: 73958288-73958289 on Build GRCh38
rs768846785 Chr.11: 73958301-73958301 on Build GRCh38
rs10793069 Chr.11: 73958289-73958289 on Build GRCh38
rs780358384 Chr.11: 73958274-73958275 on Build GRCh38
rs368168407 Chr.11: 73958547-73958547 on Build GRCh38
rs774658367 Chr.11: 73958380-73958380 on Build GRCh38
rs754357143 Chr.11: 73958431-73958431 on Build GRCh38
rs141838089 Chr.11: 73958362-73958362 on Build GRCh38
rs924617192 Chr.11: 73958599-73958599 on Build GRCh38
rs945616948 Chr.11: 73958648-73958651 on Build GRCh38
rs539106269 Chr.11: 73958331-73958331 on Build GRCh38
rs371804623 Chr.11: 73958433-73958433 on Build GRCh38
rs934015197 Chr.11: 73958260-73958260 on Build GRCh38
rs761304008 Chr.11: 73958423-73958423 on Build GRCh38
rs552505276 Chr.11: 73958632-73958632 on Build GRCh38
rs547164979 Chr.11: 73958364-73958364 on Build GRCh38
rs10793068 Chr.11: 73958265-73958265 on Build GRCh38
rs557809242 Chr.11: 73958355-73958355 on Build GRCh38
rs1026229822 Chr.11: 73958358-73958358 on Build GRCh38
rs200386171 Chr.11: 73958335-73958335 on Build GRCh38
rs141051090 Chr.11: 73958470-73958476 on Build GRCh38
rs908402184 Chr.11: 73958279-73958279 on Build GRCh38
rs777186025 Chr.11: 73958344-73958344 on Build GRCh38
rs928955488 Chr.11: 73958711-73958711 on Build GRCh38
rs936221620 Chr.11: 73958606-73958606 on Build GRCh38
rs748845872 Chr.11: 73958285-73958285 on Build GRCh38
rs375190890 Chr.11: 73958277-73958277 on Build GRCh38
rs964592090 Chr.11: 73958445-73958445 on Build GRCh38
rs768988362 Chr.11: 73958289-73958290 on Build GRCh38
rs1045877370 Chr.11: 73958690-73958690 on Build GRCh38
rs765600089 Chr.11: 73958469-73958470 on Build GRCh38
rs189302339 Chr.11: 73958707-73958707 on Build GRCh38
rs754242716 Chr.11: 73958324-73958324 on Build GRCh38
rs145187500 Chr.11: 73958360-73958360 on Build GRCh38
rs199547235 Chr.11: 73958470-73958470 on Build GRCh38
rs550129367 Chr.11: 73958551-73958551 on Build GRCh38
rs982745298 Chr.11: 73958564-73958564 on Build GRCh38
rs572611825 Chr.11: 73958638-73958638 on Build GRCh38
rs948939562 Chr.11: 73958686-73958686 on Build GRCh38
rs921311298 Chr.11: 73958604-73958604 on Build GRCh38
rs977431117 Chr.11: 73958585-73958585 on Build GRCh38
rs868366411 Chr.11: 73958654-73958654 on Build GRCh38
rs761986780 Chr.11: 73958308-73958308 on Build GRCh38
rs373170178 Chr.11: 73958472-73958472 on Build GRCh38
rs144741417 Chr.11: 73958354-73958354 on Build GRCh38
rs971708567 Chr.11: 73958534-73958534 on Build GRCh38
rs768850963 Chr.11: 73958436-73958436 on Build GRCh38
rs777544223 Chr.11: 73958475-73958475 on Build GRCh38
rs770446491 Chr.11: 73958391-73958391 on Build GRCh38

View Full Product Details