Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75262876-75263368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRB1
Entrez Gene ID
408
Gene Name
arrestin beta 1
Gene Aliases
ARB1, ARR1
UniGene
Hs.503284
Gene Chromosome Location
Chr.11: 75260122-75351831 on Build GRCh38
Transcripts
NM_004041.4 NM_020251.3 XM_011545034.2 XM_011545035.2 XM_017017750.1 XM_017017751.1 XM_017017752.1 XM_017017753.1 XM_017017754.1

SNP Detail

SNP ID Chromosome Location
rs957865151 Chr.11: 75262978-75262978 on Build GRCh38
rs1005366013 Chr.11: 75263155-75263155 on Build GRCh38
rs113528497 Chr.11: 75263127-75263127 on Build GRCh38
rs963510218 Chr.11: 75263185-75263191 on Build GRCh38
rs966404644 Chr.11: 75263205-75263205 on Build GRCh38
rs571751799 Chr.11: 75263126-75263126 on Build GRCh38
rs987186483 Chr.11: 75263211-75263211 on Build GRCh38
rs373321533 Chr.11: 75263124-75263125 on Build GRCh38
rs780842798 Chr.11: 75263191-75263191 on Build GRCh38
rs945777034 Chr.11: 75263049-75263049 on Build GRCh38
rs534168526 Chr.11: 75263329-75263329 on Build GRCh38
rs150505001 Chr.11: 75263314-75263314 on Build GRCh38
rs117788237 Chr.11: 75263030-75263030 on Build GRCh38
rs766725470 Chr.11: 75263035-75263035 on Build GRCh38
rs571591993 Chr.11: 75263170-75263170 on Build GRCh38
rs751786307 Chr.11: 75263347-75263347 on Build GRCh38
rs951981500 Chr.11: 75263297-75263297 on Build GRCh38
rs1020625416 Chr.11: 75263195-75263195 on Build GRCh38
rs35840447 Chr.11: 75263159-75263160 on Build GRCh38
rs1027923266 Chr.11: 75263282-75263282 on Build GRCh38
rs757673508 Chr.11: 75262957-75262957 on Build GRCh38
rs1047700521 Chr.11: 75263040-75263040 on Build GRCh38
rs911724557 Chr.11: 75263007-75263007 on Build GRCh38
rs183246733 Chr.11: 75263082-75263082 on Build GRCh38
rs1029209662 Chr.11: 75263200-75263200 on Build GRCh38
rs143088683 Chr.11: 75263291-75263291 on Build GRCh38
rs964829662 Chr.11: 75263268-75263268 on Build GRCh38
rs1018670226 Chr.11: 75263250-75263250 on Build GRCh38
rs937279628 Chr.11: 75263095-75263095 on Build GRCh38
rs750921446 Chr.11: 75263006-75263006 on Build GRCh38
rs116477105 Chr.11: 75262992-75262992 on Build GRCh38
rs768238874 Chr.11: 75263153-75263153 on Build GRCh38
rs907911923 Chr.11: 75263046-75263046 on Build GRCh38
rs1033227951 Chr.11: 75262964-75262964 on Build GRCh38
rs964536343 Chr.11: 75263009-75263009 on Build GRCh38
rs1013639716 Chr.11: 75262926-75262926 on Build GRCh38
rs950000763 Chr.11: 75263340-75263341 on Build GRCh38
rs552791254 Chr.11: 75263093-75263093 on Build GRCh38
rs895128591 Chr.11: 75262923-75262923 on Build GRCh38
rs146375593 Chr.11: 75263011-75263011 on Build GRCh38
rs1016837188 Chr.11: 75262983-75262983 on Build GRCh38
rs528433388 Chr.11: 75263285-75263286 on Build GRCh38
rs966432229 Chr.11: 75262998-75262998 on Build GRCh38
rs538930062 Chr.11: 75262903-75262903 on Build GRCh38
rs974577797 Chr.11: 75263269-75263269 on Build GRCh38
rs375951510 Chr.11: 75263240-75263240 on Build GRCh38
rs1042922947 Chr.11: 75263058-75263058 on Build GRCh38
rs531899328 Chr.11: 75263158-75263158 on Build GRCh38
rs564772067 Chr.11: 75262913-75262913 on Build GRCh38
rs1001678245 Chr.11: 75263147-75263147 on Build GRCh38
rs1045865364 Chr.11: 75262936-75262936 on Build GRCh38
rs575173834 Chr.11: 75262953-75262953 on Build GRCh38
rs974988222 Chr.11: 75263012-75263012 on Build GRCh38
rs560713459 Chr.11: 75262993-75262993 on Build GRCh38
rs912875408 Chr.11: 75263236-75263236 on Build GRCh38
rs371211193 Chr.11: 75263213-75263213 on Build GRCh38
rs895776783 Chr.11: 75263331-75263331 on Build GRCh38
rs560263879 Chr.11: 75263081-75263081 on Build GRCh38

View Full Product Details