Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75263270-75263778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRB1
Entrez Gene ID
408
Gene Name
arrestin beta 1
Gene Aliases
ARB1, ARR1
UniGene
Hs.503284
Gene Chromosome Location
Chr.11: 75260122-75351831 on Build GRCh38
Transcripts
NM_004041.4 NM_020251.3 XM_011545034.2 XM_011545035.2 XM_017017750.1 XM_017017751.1 XM_017017752.1 XM_017017753.1 XM_017017754.1

SNP Detail

SNP ID Chromosome Location
rs991315953 Chr.11: 75263582-75263584 on Build GRCh38
rs149680951 Chr.11: 75263406-75263406 on Build GRCh38
rs139604424 Chr.11: 75263436-75263436 on Build GRCh38
rs907880687 Chr.11: 75263358-75263358 on Build GRCh38
rs950017053 Chr.11: 75263363-75263363 on Build GRCh38
rs200753370 Chr.11: 75263741-75263741 on Build GRCh38
rs917068318 Chr.11: 75263628-75263628 on Build GRCh38
rs937429143 Chr.11: 75263661-75263661 on Build GRCh38
rs946278249 Chr.11: 75263408-75263408 on Build GRCh38
rs771469187 Chr.11: 75263582-75263582 on Build GRCh38
rs575137056 Chr.11: 75263461-75263461 on Build GRCh38
rs940648347 Chr.11: 75263744-75263744 on Build GRCh38
rs534168526 Chr.11: 75263329-75263329 on Build GRCh38
rs150505001 Chr.11: 75263314-75263314 on Build GRCh38
rs967420978 Chr.11: 75263538-75263538 on Build GRCh38
rs770781399 Chr.11: 75263547-75263547 on Build GRCh38
rs751786307 Chr.11: 75263347-75263347 on Build GRCh38
rs762947653 Chr.11: 75263477-75263477 on Build GRCh38
rs775657056 Chr.11: 75263626-75263626 on Build GRCh38
rs951981500 Chr.11: 75263297-75263297 on Build GRCh38
rs981385262 Chr.11: 75263625-75263625 on Build GRCh38
rs143088683 Chr.11: 75263291-75263291 on Build GRCh38
rs1018639329 Chr.11: 75263448-75263448 on Build GRCh38
rs954969179 Chr.11: 75263487-75263487 on Build GRCh38
rs759720270 Chr.11: 75263600-75263600 on Build GRCh38
rs950022288 Chr.11: 75263636-75263636 on Build GRCh38
rs772925752 Chr.11: 75263454-75263454 on Build GRCh38
rs981417956 Chr.11: 75263366-75263366 on Build GRCh38
rs145232974 Chr.11: 75263502-75263502 on Build GRCh38
rs996355557 Chr.11: 75263462-75263462 on Build GRCh38
rs978545163 Chr.11: 75263548-75263548 on Build GRCh38
rs937442631 Chr.11: 75263378-75263378 on Build GRCh38
rs950000763 Chr.11: 75263340-75263341 on Build GRCh38
rs753804825 Chr.11: 75263740-75263740 on Build GRCh38
rs193054175 Chr.11: 75263571-75263571 on Build GRCh38
rs952013723 Chr.11: 75263499-75263499 on Build GRCh38
rs868595145 Chr.11: 75263741-75263741 on Build GRCh38
rs77744983 Chr.11: 75263740-75263740 on Build GRCh38
rs372061566 Chr.11: 75263586-75263586 on Build GRCh38
rs769597149 Chr.11: 75263387-75263387 on Build GRCh38
rs574667536 Chr.11: 75263637-75263637 on Build GRCh38
rs1029155735 Chr.11: 75263455-75263455 on Build GRCh38
rs914664595 Chr.11: 75263690-75263690 on Build GRCh38
rs147622645 Chr.11: 75263666-75263666 on Build GRCh38
rs3893000 Chr.11: 75263400-75263400 on Build GRCh38
rs927373867 Chr.11: 75263368-75263368 on Build GRCh38
rs1041953112 Chr.11: 75263416-75263416 on Build GRCh38
rs1054612114 Chr.11: 75263390-75263390 on Build GRCh38
rs895776783 Chr.11: 75263331-75263331 on Build GRCh38
rs188840417 Chr.11: 75263498-75263498 on Build GRCh38
rs145518846 Chr.11: 75263435-75263435 on Build GRCh38

View Full Product Details