Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77116147-77116651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN5
Entrez Gene ID
726
Gene Name
calpain 5
Gene Aliases
ADNIV, HTRA3, VRNI, nCL-3
UniGene
Hs.248153
Gene Chromosome Location
Chr.11: 77066945-77126155 on Build GRCh38
Transcripts
NM_004055.4 XM_011545225.1 XM_017018223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243624 c.915G>T p.V305V Chr.11: 77116247-77116247 on Build GRCh38
6391415 c.903G>T p.E301D Chr.11: 77116235-77116235 on Build GRCh38
4669154 c.948C>A p.T316T Chr.11: 77116280-77116280 on Build GRCh38
1316583 c.899A>T p.E300V Chr.11: 77116231-77116231 on Build GRCh38
1316584 c.900G>T p.E300D Chr.11: 77116232-77116232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781901831 Chr.11: 77116334-77116334 on Build GRCh38
rs1025627111 Chr.11: 77116553-77116553 on Build GRCh38
rs935109291 Chr.11: 77116412-77116412 on Build GRCh38
rs201223346 Chr.11: 77116221-77116221 on Build GRCh38
rs150793938 Chr.11: 77116546-77116546 on Build GRCh38
rs782229930 Chr.11: 77116294-77116294 on Build GRCh38
rs782685336 Chr.11: 77116289-77116289 on Build GRCh38
rs782607655 Chr.11: 77116354-77116354 on Build GRCh38
rs901527442 Chr.11: 77116435-77116435 on Build GRCh38
rs531286881 Chr.11: 77116618-77116618 on Build GRCh38
rs782245014 Chr.11: 77116208-77116208 on Build GRCh38
rs782402253 Chr.11: 77116316-77116316 on Build GRCh38
rs782801712 Chr.11: 77116228-77116228 on Build GRCh38
rs534203457 Chr.11: 77116260-77116260 on Build GRCh38
rs541907176 Chr.11: 77116331-77116331 on Build GRCh38
rs558706779 Chr.11: 77116352-77116352 on Build GRCh38
rs1052093450 Chr.11: 77116421-77116421 on Build GRCh38
rs1023895658 Chr.11: 77116580-77116580 on Build GRCh38
rs782765710 Chr.11: 77116323-77116323 on Build GRCh38
rs781877051 Chr.11: 77116259-77116259 on Build GRCh38
rs898579535 Chr.11: 77116524-77116524 on Build GRCh38
rs781870816 Chr.11: 77116280-77116280 on Build GRCh38
rs888917049 Chr.11: 77116505-77116505 on Build GRCh38
rs781953677 Chr.11: 77116177-77116178 on Build GRCh38
rs782282556 Chr.11: 77116290-77116290 on Build GRCh38
rs782574047 Chr.11: 77116175-77116175 on Build GRCh38
rs141763172 Chr.11: 77116517-77116517 on Build GRCh38
rs782495056 Chr.11: 77116281-77116281 on Build GRCh38
rs781952011 Chr.11: 77116327-77116327 on Build GRCh38
rs371829159 Chr.11: 77116563-77116563 on Build GRCh38
rs782145328 Chr.11: 77116339-77116339 on Build GRCh38
rs781929811 Chr.11: 77116321-77116321 on Build GRCh38
rs545510595 Chr.11: 77116229-77116229 on Build GRCh38
rs369928182 Chr.11: 77116182-77116182 on Build GRCh38
rs782018269 Chr.11: 77116220-77116220 on Build GRCh38
rs782346799 Chr.11: 77116300-77116300 on Build GRCh38
rs782142591 Chr.11: 77116231-77116231 on Build GRCh38
rs367696171 Chr.11: 77116555-77116555 on Build GRCh38
rs782006156 Chr.11: 77116309-77116309 on Build GRCh38
rs781786380 Chr.11: 77116248-77116248 on Build GRCh38
rs1050782109 Chr.11: 77116449-77116449 on Build GRCh38
rs376572382 Chr.11: 77116351-77116351 on Build GRCh38
rs548227479 Chr.11: 77116621-77116621 on Build GRCh38
rs759951214 Chr.11: 77116172-77116172 on Build GRCh38
rs969667449 Chr.11: 77116614-77116614 on Build GRCh38
rs561867610 Chr.11: 77116361-77116361 on Build GRCh38
rs369500943 Chr.11: 77116293-77116293 on Build GRCh38
rs199886178 Chr.11: 77116223-77116223 on Build GRCh38
rs376987414 Chr.11: 77116326-77116326 on Build GRCh38
rs997869775 Chr.11: 77116215-77116215 on Build GRCh38
rs11606456 Chr.11: 77116177-77116177 on Build GRCh38
rs782454728 Chr.11: 77116292-77116292 on Build GRCh38
rs533231666 Chr.11: 77116450-77116450 on Build GRCh38
rs1006014569 Chr.11: 77116507-77116507 on Build GRCh38
rs11604536 Chr.11: 77116314-77116314 on Build GRCh38
rs782400642 Chr.11: 77116579-77116579 on Build GRCh38
rs372926639 Chr.11: 77116349-77116349 on Build GRCh38
rs782709664 Chr.11: 77116237-77116237 on Build GRCh38
rs577452164 Chr.11: 77116226-77116226 on Build GRCh38
rs782716849 Chr.11: 77116270-77116270 on Build GRCh38
rs564257332 Chr.11: 77116447-77116447 on Build GRCh38
rs918617817 Chr.11: 77116301-77116301 on Build GRCh38
rs782775407 Chr.11: 77116333-77116333 on Build GRCh38
rs370028458 Chr.11: 77116209-77116209 on Build GRCh38
rs541334955 Chr.11: 77116431-77116431 on Build GRCh38
rs374973991 Chr.11: 77116210-77116210 on Build GRCh38
rs527456562 Chr.11: 77116372-77116372 on Build GRCh38
rs782030609 Chr.11: 77116622-77116622 on Build GRCh38

View Full Product Details